Merck
Increasing nontuberculous mycobacteria infection in cystic fibrosis.
Journal of cystic fibrosis : official journal of the European Cystic Fibrosis Society (2014-06-12)
Ophir Bar-On, Huda Mussaffi, Meir Mei-Zahav, Dario Prais, Guy Steuer, Patrick Stafler, Shai Hananya, Hannah Blau
ZUSAMMENFASSUNG
Nontuberculous mycobacteria (NTM) are emerging infections in the CF population. To assess NTM infection prevalence and associated features in our CF clinic population. Patient records, 2002-2011, were reviewed for NTM infection. FEV1, pancreatic function, sputum microbiology, and serum cytokines were compared in patients with and without NTM infection. Incidence rate of NTM infection increased from 0 in 2002 to 8.7% in 2011 (p<0.001). NTM infection prevalence increased 3-fold from 5% (4/79) in 2003 to 14.5% (16/110) in 2011 (p=0.05). Prevalence of chronic NTM lung disease has decreased somewhat since a peak in 2009, with institution of aggressive triple therapy. Of NTM-infected compared to uninfected patients, 88.2% vs. 60.3% had a known 'severe' CFTR genotype (p=0.04), 88.2% vs. 58.9% were pancreatic insufficient (p=0.02); 70.6% vs. 43.8% had chronic Pseudomonas aeruginosa (p=0.06); 75% vs. 32% had Aspergillus infection (p=0.007) and 23.5% vs 2.7% had allergic bronchopulmonary aspergillosis (p=0.01). Patients infected with Mycobacterium abscessus had increased TGF-β, TNF-α, IL-1β, IL-2, IL-4 and IL-5 levels (p<0.05). There was no difference in cytokine levels for all NTM infected compared to uninfected patients. M. abscessus comprised 46% of all NTM infections. Comparing M. abscessus versus other NTM, duration was 10.5 (1-118) months versus 1 (1-70) month, median (range) (p=0.004); lung disease occurred in 69% versus 17% (p=0.0004), with sputum conversion in 4/11 versus 5/6, respectively (NS). NTM incidence and prevalence have increased dramatically in our CF clinic, associated with a severe CF genotype and phenotype. M. abscessus, the most prevalent NTM, caused prolonged infection despite therapy. There has been some decrease in the prevalence of NTM lung disease since 2009.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
Glycerin, analytical standard
Sigma-Aldrich
Glycerin, tested according to Ph. Eur., anhydrous