Merck
  • Effects of UV exclusion on the physiology and phenolic composition of leaves and berries of Vitis vinifera cv. Graciano.
Effects of UV exclusion on the physiology and phenolic composition of leaves and berries of Vitis vinifera cv. Graciano.
Journal of the science of food and agriculture (2014-05-14)
María-Ángeles Del-Castillo-Alonso, María P Diago, Laura Monforte, Javier Tardaguila, Javier Martínez-Abaigar, Encarnación Núñez-Olivera
ZUSAMMENFASSUNG
Ultraviolet (UV) radiation induces adaptive responses that can be used for plant production improvement. The aim of this study was to assess the effect of solar UV exclusion on the physiology and phenolic compounds of leaves and berry skins of Vitis vinifera L. cv. Graciano under field conditions. Phenolic compounds were analyzed globally and individually in both the vacuolar fraction and, for the first time in grapevine, the cell wall-bound fraction. These different locations may represent diverse modalities of phenolic response to and protection against UV. UV exclusion led to a decrease in Fv /Fm in leaves, revealing that solar UV is needed for adequate photoprotection. Only p-caffeoyl-tartaric acid from the soluble fraction of leaves and myricetin-3-O-glucoside from the soluble fraction of berry skins were significantly higher in the presence of UV radiation, and thus they could play a role in UV protection. Other hydroxycinnamic acids, flavonols, flavanols and stilbenes did not respond to UV exclusion. UV exclusion led to subtle changes in leaves and berry skins of Graciano cultivar, which would be well adapted to current UV levels. This may help support decision-making on viticultural practices modifying UV exposure of leaves and berries, which could improve grape and wine quality.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Calciumcarbonat, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Chlorophyll a, from spinach
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Calciumcarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Chlorophyll a, from Anacystis nidulans algae
Supelco
Aceton, analytical standard
Sigma-Aldrich
trans-Ferulasäure, 99%
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Supelco
Calciumcarbonat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Calciumcarbonat, ReagentPlus®