Merck
Evaluation of a dansyl-based amino acid DNSBA as an imaging probe for apoptosis detection.
Apoptosis : an international journal on programmed cell death (2015-01-19)
Min Tang, Jiaguo Huang, Xinxian Weng, Lifang Yang, Meihui Liu, Ming Zhou, Xiaobo Wang, Jinghe Gao, Wei Yi, Wenbin Zeng, Lunquan Sun, Ya Cao
ZUSAMMENFASSUNG
Imaging agents that enable direct detection of apoptosis are highly desirable in the field of monitoring chemotherapeutic response as well as early diagnosis and disease monitoring. Previous work demonstrated that the dansyled amino acid DNSBA is used to specifically and selectively detect apoptotic cancer cells at the both early and late stages, but the mechanism remains unclear. In this work, we evaluated DNSBA as a tool for monitoring cell apoptosis in CNE1 tumor cell models both in vitro and ex vivo after its in vivo administration, which was confirmed by other assays. The ability of DNSBA to detect multiple pathways and different stages of apoptosis leading to cell death may be advantageous in the evaluation of cancer treatment indicative of a positive therapeutic outcome. The uptake change of molecular probes DNSBA in CNE1 cells represented the changes of apoptotic rate in a caspase-dependent manner. However, the accumulation of DNSBA in apoptotic cells did not increase with the enhanced membrane permeability. Furthermore, ex vivo study demonstrated DNSBA has a similar pattern as the TUNEL-positive cells. In conclusion, DNSBA cellular imaging is useful for the early assessment of treatment-induced apoptosis, and thus may act as a substitute for Annexin V for assessing treatment response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
N-Bromsuccinimid, ReagentPlus®, 99%
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Adenosin-5′-diphosphoribose Natriumsalz, ≥93%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, tested according to Ph. Eur.