Merck
  • miR-200a suppresses cell growth and migration by targeting MACC1 and predicts prognosis in hepatocellular carcinoma.
miR-200a suppresses cell growth and migration by targeting MACC1 and predicts prognosis in hepatocellular carcinoma.
Oncology reports (2014-12-09)
Jiye Feng, Jinbo Wang, Mingliang Chen, Gun Chen, Zongyang Wu, Liping Ying, Qifeng Zhuo, Jianlei Zhang, Weilin Wang
ZUSAMMENFASSUNG
miR-200a suppresses tumor development and progression; however, its role in tumor growth and metastasis of hepatocellular carcinoma (HCC) and the underlying mechanism have not been elucidated. In the present study, we identified that miR-200a was markedly downregulated in HCC and exerted suppressive effects on tumor cell growth and metastasis. We identified that miR-200a suppressed tumor growth and metastasis by directly targeting MACC1. In addition, HCC patients with low miR-200a expression had significantly worse prognosis than those with high expression of miR-200a. These findings suggest that miR-200a may be recognized as a novel potential biomarker to predict the survival of patients with HCCs following liver transplantation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Maleinsäure, ReagentPlus®, ≥99% (HPLC)
Supelco
Maleinsäure, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Natriumzitrat, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Maleinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Maleinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioUltra, Molecular Biology, 1 M in H2O
Supelco
Digoxigenin, analytical standard
Digoxigenin, European Pharmacopoeia (EP) Reference Standard