Merck
  • Defective functionality of HDL particles in familial apoA-I deficiency: relevance of alterations in HDL lipidome and proteome.
Defective functionality of HDL particles in familial apoA-I deficiency: relevance of alterations in HDL lipidome and proteome.
Journal of lipid research (2014-10-25)
Fabiana Rached, Raul D Santos, Laurent Camont, Marcio H Miname, Marie Lhomme, Carolane Dauteuille, Sora Lecocq, Carlos V Serrano, M John Chapman, Anatol Kontush
ZUSAMMENFASSUNG
To evaluate functional and compositional properties of HDL in subjects from a kindred of genetic apoA-I deficiency, two homozygotes and six heterozygotes, with a nonsense mutation at APOA1 codon -2, Q[-2]X, were recruited together with age- and sex-matched healthy controls (n = 11). Homozygotes displayed undetectable plasma levels of apoA-I and reduced levels of HDL-cholesterol (HDL-C) and apoC-III (5.4% and 42.6% of controls, respectively). Heterozygotes displayed low HDL-C (21 ± 9 mg/dl), low apoA-I (79 ± 24 mg/dl), normal LDL-cholesterol (132 ± 25 mg/dl), and elevated TG (130 ± 45 mg/dl) levels. Cholesterol efflux capacity of ultracentrifugally isolated HDL subpopulations was reduced (up to -25%, P < 0.01, on a glycerophospholipid [GP] basis) in heterozygotes versus controls. Small, dense HDL3 and total HDL from heterozygotes exhibited diminished antioxidative activity (up to -48%, P < 0.001 on a total mass basis) versus controls. HDL subpopulations from both homozygotes and heterozygotes displayed altered chemical composition, with depletion in apoA-I, GP, and cholesteryl ester; enrichment in apoA-II, free cholesterol, and TG; and altered phosphosphingolipidome. The defective atheroprotective activities of HDL were correlated with altered lipid and apo composition. These data reveal that atheroprotective activities of HDL particles are impaired in homozygous and heterozygous apoA-I deficiency and are intimately related to marked alterations in protein and lipid composition.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Supelco
Ammoniumformat, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammoniumformat, reagent grade, 97%
Sigma-Aldrich
Ammoniumformat, ≥99.995% trace metals basis
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Ammoniumformiat -Lösung, BioUltra, 10 M in H2O
Sigma-Aldrich
L-Serin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
2-Aminoethyl-dihydrogen-phosphat
Sigma-Aldrich
Ammoniumformat, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
1,2-Distearoyl-sn-Glycero-3-Phosphocholin, ≥99%
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
1,2-Dipalmitoyl-sn-Glycero-3-Phosphocholin aus Staphylococcus aureus, ≥99% (TLC)
Sigma-Aldrich
1,2-Dipalmitoyl-sn-Glycero-3-Phosphocholin aus Staphylococcus aureus, semisynthetic, ≥99%
Sigma-Aldrich
2-Oleoyl-1-Palmitoyl-sn-Glycero-3-Phosphocholin, ≥99.0% (TLC)
Sigma-Aldrich
L-Serin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
1,2-Dimyristoyl-sn-Glycero-3-Phosphocholin, ≥99%
Supelco
L-Serin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O
Sigma-Aldrich
2-Oleoyl-1-Palmitoyl-sn-Glycero-3-Phosphocholin, ≥95.5% (GC), ≥98% (TLC)
Supelco
L-Serin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland