Merck
  • Spatial and temporal distribution of chiral pesticides in Calanus spp. from three Arctic fjords.
Spatial and temporal distribution of chiral pesticides in Calanus spp. from three Arctic fjords.
Environmental pollution (Barking, Essex : 1987) (2014-06-22)
Pernilla Carlsson, Nicholas A Warner, Ingeborg G Hallanger, Dorte Herzke, Roland Kallenborn
ZUSAMMENFASSUNG
Concentration and enantiomeric fractions (EFs) of chiral chlorinated pesticides (α-hexachlorocyclohexane (α-HCH), trans-, cis- and oxychlordane) were determined in Arctic zooplankton, mainly Calanus spp. collected in the period 2007-11 from Svalbard fjords and open pack-ice. The temporal and spatial enantiomer distribution varied considerably for all species and chiral pesticides investigated. An overall enantiomeric excess of (+)-oxychlordane (EF 0.53-0.86) were observed. Cis-chlordane was close to racemic (EF 0.46-0.55), while EF for trans-chlordane varied between 0.29 and 0.55, and between 0.38 and 0.59 for α-HCH. The biodegradation potential for trans-chlordane was higher compared to cis-chlordane. The comprehensive statistical evaluation of the data set revealed that the EF distribution of α-HCH was affected by ice cover to a higher extent compared to cis-chlordane. Potential impact from benthic processes on EFs in zooplankton is an interesting feature and should be further investigated. Enantiomeric selective analyses may be a suitable tool for investigations of climate change related influences on Arctic ecosystems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar