Merck
  • Oxidative stress induces phosphorylation of neuronal NOS in cardiomyocytes through AMP-activated protein kinase (AMPK).
Oxidative stress induces phosphorylation of neuronal NOS in cardiomyocytes through AMP-activated protein kinase (AMPK).
Biochemical and biophysical research communications (2015-03-04)
Rekha Kar, Dean L Kellogg, Linda J Roman
ZUSAMMENFASSUNG
Neuronal nitric oxide synthase (nNOS) plays a critical role in regulating cardiomyocyte function. nNOS was reported to decrease superoxide production in the myocardium by inhibiting the function of xanthine oxidoreductase. However, the effect of oxidative stress on nNOS in cardiomyocytes has not been determined. We report here that brief exposure of HL-1 cardiomyocytes to hydrogen peroxide (H2O2) induces phosphorylation of nNOS at serine 1412. This increase in phosphorylation was concomitant with increased nitric oxide (NO) production. Prolonged exposure to the oxidant, however, resulted in decreased expression of the protein. H2O2 treatment for short periods also stimulated phosphorylation of AKT and AMPK. H2O2-induced phosphorylation of nNOS was reduced when AMPK activity was inhibited by compound C, suggesting that AMPK is a mediator of oxidative stress-induced phosphorylation of nNOS. However, inhibition of AKT activity by the pan AKT inhibitor, AKTi, had no effect on nNOS phosphorylation caused by H2O2. These data demonstrate the novel regulation of nNOS phosphorylation and expression by oxidative stress.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Threonin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonin, reagent grade, ≥98% (HPLC)
SAFC
L-Threonin
Sigma-Aldrich
L-Threonin, BioXtra, ≥99.5% (NT)
Supelco
L-Threonin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Serin, ≥98% (TLC)
Sigma-Aldrich
DL-Serin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Serin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Threonin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
L-Threonin, European Pharmacopoeia (EP) Reference Standard