Merck
  • High red blood cell nitric oxide synthase activation is not associated with improved vascular function and red blood cell deformability in sickle cell anaemia.
High red blood cell nitric oxide synthase activation is not associated with improved vascular function and red blood cell deformability in sickle cell anaemia.
British journal of haematology (2014-10-16)
Marijke Grau, Anaïs Mozar, Keyne Charlot, Yann Lamarre, Linda Weyel, Frank Suhr, Bianca Collins, Stéphane Jumet, Marie-Dominique Hardy-Dessources, Marc Romana, Nathalie Lemonne, Maryse Etienne-Julan, Sophie Antoine-Jonville, Wilhelm Bloch, Philippe Connes
ZUSAMMENFASSUNG
Human red blood cells (RBC) express an active and functional endothelial-like nitric oxide (NO) synthase (RBC-NOS). We report studies on RBC-NOS activity in sickle cell anaemia (SCA), a genetic disease characterized by decreased RBC deformability and vascular dysfunction. Total RBC-NOS content was not significantly different in SCA patients compared to healthy controls; however, using phosphorylated RBC-NOS-Ser(1177) as a marker, RBC-NOS activation was higher in SCA patients as a consequence of the greater activation of Akt (phosphorylated Akt-Ser(473) ). The higher RBC-NOS activation in SCA led to higher levels of S-nitrosylated α- and β-spectrins, and greater RBC nitrite and nitrotyrosine levels compared to healthy controls. Plasma nitrite content was not different between the two groups. Laser Doppler flowmetric experiments demonstrated blunted microcirculatory NO-dependent response under hyperthermia in SCA patients. RBC deformability, measured by ektacytometry, was reduced in SCA in contrast to healthy individuals, and pre-shearing RBC in vitro did not improve deformability despite an increase of RBC-NOS activation. RBC-NOS activation is high in freshly drawn blood from SCA patients, resulting in high amounts of NO produced by RBC. However, this does not result in improved RBC deformability and vascular function: higher RBC-NO is not sufficient to counterbalance the enhanced oxidative stress in SCA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Tricin, ≥99% (titration)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
L-Threonin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Threonin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
SAFC
L-Threonin
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Tricin, BioPerformance Certified, suitable for cell culture, ≥99% (titration)
Sigma-Aldrich
Tricin, BioXtra, pH 4.0-6.0 (1 M in H2O), ≥99% (titration)
Sigma-Aldrich
L-Threonin, BioXtra, ≥99.5% (NT)
Supelco
L-Threonin, Pharmaceutical Secondary Standard; Certified Reference Material