Merck
Lung tumors in mice induced by "whole-life" inorganic arsenic exposure at human-relevant doses.
Archives of toxicology (2014-07-10)
Michael P Waalkes, Wei Qu, Erik J Tokar, Grace E Kissling, Darlene Dixon
ZUSAMMENFASSUNG
In mice, inorganic arsenic in the drinking water in the parts per million range via the dam during in utero life or with whole-life exposure is a multi-site carcinogen in the offspring. However, human arsenic exposure is typically in the parts per billion (ppb) range. Thus, we studied "whole-life" inorganic arsenic carcinogenesis in mice at levels more relevant to humans. Breeder male and female CD1 mice were exposed to 0, 50, 500 or 5,000 ppb arsenic (as sodium arsenite) in the drinking water for 3 weeks prior to breeding, during pregnancy and lactation, and after weaning (at week 3) groups of male and female offspring (initial n = 40) were exposed for up to 2 years. Tumors were assessed in these offspring. Arsenic exposure had no effect on pregnant dam weights or water consumption, litter size, offspring birthweight or weight at weaning compared to control. In male offspring mice, arsenic exposure increased (p < 0.05) bronchiolo-alveolar tumor (adenoma or carcinoma) incidence at 50-ppb group (51 %) and 500-ppb group (54 %), but not at 5,000-ppb group (28 %) compared to control (22 %). These arsenic-induced bronchiolo-alveolar tumors included increased (p < 0.05) carcinoma at 50-ppb group (27 %) compared to controls (8 %). An increase (p < 0.05) in lung adenoma (25 %) in the 50-ppb group compared to control (11 %) occurred in female offspring. Thus, in CD1 mice whole-life arsenic exposure induced lung tumors at human-relevant doses (i.e., 50 and 500 ppb).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salpetersäure, ACS reagent, 70%
Sigma-Aldrich
Perchlorsäure, ACS reagent, 70%
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Salpetersäure, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Perchlorsäure, 70%, 99.999% trace metals basis
Sigma-Aldrich
Perchlorsäure, ACS reagent, 60%
Sigma-Aldrich
Salpetersäure, red, fuming, HNO3 >90 %
Supelco
Salpetersäure-Konzentrat, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%
Sigma-Aldrich
Arsen, 99.999% trace metals basis
Sigma-Aldrich
Arsen, powder, ≥99.997% trace metals basis
Supelco
Perchlorsäure-Konzentrat, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Salpeter-14N-Säurelösung, ~10 N in H2O, 99.99 atom % 14N