Merck
  • A comprehensive review in current developments of benzothiazole-based molecules in medicinal chemistry.
A comprehensive review in current developments of benzothiazole-based molecules in medicinal chemistry.
European journal of medicinal chemistry (2014-12-03)
Rangappa S Keri, Mahadeo R Patil, Siddappa A Patil, Srinivasa Budagumpi
ZUSAMMENFASSUNG
Benzothiazole (BTA) and its derivatives are the most important heterocyclic compounds, which are common and integral feature of a variety of natural products and pharmaceutical agents. BTA shows a variety of pharmacological properties, and its analogs offer a high degree of structural diversity that has proven useful for the search of new therapeutic agents. The broad spectrum of pharmacological activity in individual BTA derivative indicates that, this series of compounds is of an undoubted interest. The related research and developments in BTA-based medicinal chemistry have become a rapidly developing and increasingly active topic. Particularly, numerous BTA-based compounds as clinical drugs have been extensively used in practice to treat various types of diseases with high therapeutic potency. This work systematically gives a comprehensive review in current developments of BTA-based compounds in the whole range of medicinal chemistry as anticancer, antibacterial, antifungal, antiinflammatory, analgesic, anti-HIV, antioxidant, anticonvulsant, antitubercular, antidiabetic, antileishmanial, antihistaminic, antimalarial and other medicinal agents. It is believed that, this review article is helpful for new thoughts in the quest for rational designs of more active and less toxic BTA-based drugs, as well as more effective diagnostic agents and pathologic probes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Brenzcatechin, ≥99%
Sigma-Aldrich
1,2-Dihydroxybenzol, ReagentPlus®, ≥99%
Sigma-Aldrich
Brenzcatechin, purified by sublimation, ≥99.5%