Merck
  • The orphan nuclear receptor NR4A1 specifies a distinct subpopulation of quiescent myeloid-biased long-term HSCs.
The orphan nuclear receptor NR4A1 specifies a distinct subpopulation of quiescent myeloid-biased long-term HSCs.
Stem cells (Dayton, Ohio) (2014-10-07)
Ruben H Land, Anna K Rayne, Ashley N Vanderbeck, Trevor S Barlowe, Shwetha Manjunath, Matthew Gross, Sophie Eiger, Peter S Klein, Nicole R Cunningham, Jian Huang, Stephen G Emerson, Jennifer A Punt
ZUSAMMENFASSUNG
Hematopoiesis is maintained throughout life by self-renewing hematopoietic stem cells (HSCs) that differentiate to produce both myeloid and lymphoid cells. The NR4A family of orphan nuclear receptors, which regulates cell fate in many tissues, appears to play a key role in HSC proliferation and differentiation. Using a NR4A1(GFP) BAC transgenic reporter mouse we have investigated NR4A1 expression and its regulation in early hematopoiesis. We show that NR4A1 is most highly expressed in a subset of Lin(-) Sca-1(+) c-Kit(+) CD48(-) CD150(+) long-term (LT) HSCs, and its expression is tightly associated with HSC quiescence. We also show that NR4A1 expression in HSCs is induced by PGE2, a known enhancer of stem cell engraftment potential. Finally, we find that both NR4A1(GFP+) and NR4A1(GFP-) HSCs successfully engraft primary and secondary irradiated hosts; however, NR4A1(GFP+) HSCs are distinctly myeloid-biased. These results show that NR4A1 expression identifies a highly quiescent and distinct population of myeloid-biased LT-HSCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumhydrogendifluorid, reagent grade, 95%
Sigma-Aldrich
Kalium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Kaliumhydrid, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Ammoniumhydrogendifluorid, 99.999% trace metals basis
Sigma-Aldrich
Azodicarbonsäure-diethylester -Lösung, purum, ~40% in toluene (H-NMR)
Supelco
N,N′-Bis(acryloyl)cystamin, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Kaliumhydrid, in paraffin
Sigma-Aldrich
Pyronin Y, for NA electrophoresis
Sigma-Aldrich
Pyronin G, certified by the BSC, Dye content 50 %
Sigma-Aldrich
Pyronin G, suitable for microscopy