Merck
Nonmyeloablative allogeneic hematopoietic stem cell transplantation for GATA2 deficiency.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation (2014-08-12)
Jennifer Grossman, Jennifer Cuellar-Rodriguez, Juan Gea-Banacloche, Christa Zerbe, Katherine Calvo, Thomas Hughes, Fran Hakim, Kristen Cole, Mark Parta, Alexandra Freeman, Steven M Holland, Dennis D Hickstein
ZUSAMMENFASSUNG
We treated 14 patients with GATA2 deficiency using a nonmyeloablative allogeneic hematopoietic stem cell transplantation regimen. Four patients received peripheral blood stem cells from matched related donors (MRD), 4 patients received peripheral blood stem cells from matched unrelated donors (URD), 4 patients received hematopoietic stem cells from umbilical cord blood donors (UCB), and 2 patients received bone marrow cells from haploidentical related donors. MRD and URD recipients received conditioning with 3 days of fludarabine and 200 cGy total body irradiation (TBI). Haploidentical related donor recipients and UCB recipients received cyclophosphamide and 2 additional days of fludarabine along with 200 cGY TBI. MRD, URD, and UCB recipients received tacrolimus and sirolimus for post-transplantation immunosuppression, whereas haploidentical recipients received high-dose cyclophosphamide followed by tacrolimus and mycophenolate mofetil. Eight patients are alive with reconstitution of the severely deficient monocyte, B cell, and natural killer cell populations and reversal of the clinical phenotype at a median follow-up of 3.5 years. Two patients (1 URD recipient and 1 UCB recipient) rejected the donor graft and 1 MRD recipient relapsed with myelodysplastic syndrome after transplantation. We are currently using a high-dose conditioning regimen with busulfan and fludarabine in patients with GATA2 deficiency to achieve more consistent engraftment and eradication of the malignant myeloid clones.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rapamycin aus Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
FK-506 Monohydrat, ≥98% (HPLC)
USP
Tacrolimus, United States Pharmacopeia (USP) Reference Standard