Merck
  • E-selectin expression and BRAF status in papillary thyroid carcinomas: Correlation with clinicopathologic features.
E-selectin expression and BRAF status in papillary thyroid carcinomas: Correlation with clinicopathologic features.
Surgery (2014-12-03)
Paolo Miccoli, Liborio Torregrossa, Nicla Borrelli, Gabriele Materazzi, Andrea Cacciato Insilla, Mario Miccoli, Fulvio Basolo
ZUSAMMENFASSUNG
Cell adhesion molecules, represented by the immunoglobulin family and selectins, play an important role in the progression of cancer. A correlation between selectins and tumor aggressiveness has been demonstrated in several reports. Eighty-eight patients (mean age, 41.0 ± 14 years) with papillary thyroid carcinoma (conventional variant and sized approximately 20 mm) were divided in 2 groups: 41 with encapsulated tumors and 47 with tumors with extrathyroidal extension. E-selectin expression was evaluated by immunohistochemical staining and semiquantitative real-time reverse-transcription polymerase chain reaction and normalized by calculating the z-score (positive: value above the population mean; negative: below the mean). Lymph node metastasis (LNM) was found in 2 of 41 encapsulated tumors (4.8%) and in 19 of 47 tumors (40.4%) with extrathyroidal extension. BRAF mutation was present in 21 encapsulated tumors (51.2%) and in 31 tumors with extrathyroidal extension (65.9%). The mean E-selectin z-score was -0.32 for encapsulated tumors and 0.28 for tumors with extrathyroidal extension. E-selectin expression correlates with neoplastic infiltration (P = .04), the American Joint Commission on Cancer stage (P = .02), and BRAF mutation (P = .03). E-selectin overexpression in association with BRAF mutation status could promote a more aggressive phenotype in papillary thyroid carcinoma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
o-Xylen, reagent grade, ≥98.0%
Sigma-Aldrich
o-Xylen, anhydrous, 97%
Supelco
o-Xylen, analytical standard
Supelco
o-Xylen, Pharmaceutical Secondary Standard; Certified Reference Material