Merck
  • Immunophenotypic features by multiparameter flow cytometry can help distinguish low grade B-cell lymphomas with plasmacytic differentiation from plasma cell proliferative disorders with an unrelated clonal B-cell process.
Immunophenotypic features by multiparameter flow cytometry can help distinguish low grade B-cell lymphomas with plasmacytic differentiation from plasma cell proliferative disorders with an unrelated clonal B-cell process.
British journal of haematology (2015-02-04)
Flavia G Rosado, William G Morice, Rong He, Matthew T Howard, Michael Timm, Ellen D McPhail
ZUSAMMENFASSUNG
Highly sensitive flow cytometry studies may incidentally identify B cell clones when used to assess plasma cell clonality in bone marrows. Clinical history, which can help differentiate related clones (low grade B cell lymphoma with plasmacytic differentiation/LBCL-PD) from unrelated ones (plasma cell proliferative disorder (PCPD) with an unrelated B cell clone), is often unavailable in referred specimens. We sought to identify morphologic or phenotypic features that would help predict the significance of these clones in the absence of history. We included only cases with identical light chain B and plasma cell clones, as determined by 6-color flow cytometry with additional DNA ploidy analysis, in which the relationship between clones could be established by review of medical records. There were 26 cases; 18 were related (14 were Waldenstrom macroglobulinemia) and eight were unrelated (seven multiple myeloma). Features seen exclusively in LBCL-PD include CD19+/CD45+ clonal plasma cell phenotype (66·7%, P = 0·0022) and morphologic features such as paratrabecular bone marrow involvement, increased mast cells, and plasma cells surrounding B-cell nodules. Aneuploidy was identified exclusively in PCPD cases (75%, P = 0·000028). We conclude that CD19+/CD45+ clonal plasma cell phenotype and aneuploidy are useful in distinguishing related clones (LBCL-PD) from unrelated clones (PCPD).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ammoniumchlorid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammoniumchlorid, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammoniumchlorid, 99.998% trace metals basis
Sigma-Aldrich
Aphidicolin, ≥98% (HPLC), powder
Supelco
Ammoniumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammoniumchlorid, 99.99% trace metals basis
Sigma-Aldrich
Ammoniumchlorid, BioUltra, ≥99.5% (AT)
Supelco
Ammonium-Ionenlösung für ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Ammoniumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium-14N-chlorid, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Supelco
Aphidicolin, analytical standard