Merck
  • Sustained alterations of hypothalamic tanycytes during posttraumatic hypopituitarism in male mice.
Sustained alterations of hypothalamic tanycytes during posttraumatic hypopituitarism in male mice.
Endocrinology (2014-03-08)
Guillaume Osterstock, Taoufik El Yandouzi, Nicola Romanò, Danielle Carmignac, Fanny Langlet, Nathalie Coutry, Anne Guillou, Marie Schaeffer, Norbert Chauvet, Charlotte Vanacker, Evelyne Galibert, Bénédicte Dehouck, Iain C A F Robinson, Vincent Prévot, Patrice Mollard, Nikolaus Plesnila, Pierre-François Méry
ZUSAMMENFASSUNG
Traumatic brain injury is a leading cause of hypopituitarism, which compromises patients' recovery, quality of life, and life span. To date, there are no means other than standardized animal studies to provide insights into the mechanisms of posttraumatic hypopituitarism. We have found that GH levels were impaired after inducing a controlled cortical impact (CCI) in mice. Furthermore, GHRH stimulation enhanced GH to lower level in injured than in control or sham mice. Because many characteristics were unchanged in the pituitary glands of CCI mice, we looked for changes at the hypothalamic level. Hypertrophied astrocytes were seen both within the arcuate nucleus and the median eminence, two pivotal structures of the GH axis, spatially remote to the injury site. In the arcuate nucleus, GHRH neurons were unaltered. In the median eminence, injured mice exhibited unexpected alterations. First, the distributions of claudin-1 and zonula occludens-1 between tanycytes were disorganized, suggesting tight junction disruptions. Second, endogenous IgG was increased in the vicinity of the third ventricle, suggesting abnormal barrier properties after CCI. Third, intracerebroventricular injection of a fluorescent-dextran derivative highly stained the hypothalamic parenchyma only after CCI, demonstrating an increased permeability of the third ventricle edges. This alteration of the third ventricle might jeopardize the communication between the hypothalamus and the pituitary gland. In conclusion, the phenotype of CCI mice had similarities to the posttraumatic hypopituitarism seen in humans with intact pituitary gland and pituitary stalk. It is the first report of a pathological status in which tanycyte dysfunctions appear as a major acquired syndrome.