Merck
  • Protective effects of lactococci strains delivering either IL-10 protein or cDNA in a TNBS-induced chronic colitis model.
Protective effects of lactococci strains delivering either IL-10 protein or cDNA in a TNBS-induced chronic colitis model.
Journal of clinical gastroenterology (2014-10-08)
Silvina del Carmen, Rebeca Martín Rosique, Tessália Saraiva, Meritxell Zurita-Turk, Anderson Miyoshi, Vasco Azevedo, Alejandra de Moreno de LeBlanc, Philippe Langella, Luis G Bermúdez-Humarán, Jean Guy LeBlanc
ZUSAMMENFASSUNG
Oral treatment with Lactococcus lactis strains secreting the anti-inflammatory cytokine interleukin (IL)-10 has previously shown success as a therapy for inflammatory bowel diseases (IBD). Our aim was to compare the protective effects of IL-10, delivered by recombinant lactoccoci using 2 novel expression systems, in a murine colitis model mimicking the relapsing nature of IBD. The first system is based on a Stress-Inducible Controlled Expression system for the production and delivery of heterologous proteins at mucosal surfaces and the second allows the delivery to the host cells of an il-10 cDNA cassette, harbored in a eukaryotic DNA expression vector (pValac). Colitis was induced in female BALB/c mice by intrarectal injection of 2,4,6-trinitrobenzenesulphonic acid (TNBS). Mice that recovered received one of the bacteria treatments or saline solution orally during 14 days. Colitis was reactivated 25 days after the first TNBS injection with a second TNBS challenge. Three days after colitis reactivation, cytokine profiles and inflammation in colon samples were evaluated. Animals (N=9) receiving L. lactis strains secreting IL-10 using Stress-Inducible Controlled Expression system or delivering pValac:il-10 plasmid showed lower weight loss (P<0.005), lower damage scores (P<0.005), and immune activation in their large intestines compared with inflamed nontreated mice. Our results confirm the protective effect of IL-10 delivered either as a protein or as a cDNA in a colitis model mimicking the relapsing nature of IBD and provides a step further in the "proof-of-concept" of genetically engineered bacteria as a valid system to deliver therapeutic molecules at mucosal level.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Picrylsulfonsäure -Lösung, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Xylazin -hydrochlorid, ≥99.0% (HPLC)
Sigma-Aldrich
Picrylsulfonsäure -Lösung, 1 M in H2O
Xylazin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard