Merck
VGLL1 expression is associated with a triple-negative basal-like phenotype in breast cancer.
Endocrine-related cancer (2014-06-04)
María Ángeles Castilla, María Ángeles López-García, María Reina Atienza, Juan Manuel Rosa-Rosa, Juan Díaz-Martín, María Luisa Pecero, Begoña Vieites, Laura Romero-Pérez, Javier Benítez, Annarica Calcabrini, José Palacios
ZUSAMMENFASSUNG
Vestigial-like 1 (VGLL1) is a poorly characterized gene encoding a transcriptional co-activator structurally homologous to TAZ and YAP that modulates the Hippo pathway in Drosophila. In this study, we examined the expression of VGLL1 and its intronic miRNA, miR-934, in breast cancer. VGLL1 and miR-934 expression miRNA profiling was carried out on frozen samples of grade 3 invasive ductal carcinomas. VGLL1 protein was also examined in 433 sporadic and BRCA1-associated breast carcinomas on tissue microarrays. RNA-seq data from The Cancer Genome Atlas (TCGA) was used to confirm differences in VGLL1 and miR-934 expression in different breast cancer subtypes, and to correlate their expression with that of other genes and miRNAs. Of 28 miRNAs differentially expressed in estrogen receptor (ER)-positive and ER-negative grade 3 breast carcinomas, miR-934 was most strongly upregulated in ER-negative carcinomas, and its expression was correlated with that of VGLL1. Nuclear VGLL1 expression was observed in 13% of sporadic breast carcinomas, and while VGLL1 was only occasionally found in luminal A (0.70%) and B (5.60%) carcinomas, it was often expressed in HER2-positive (17%), triple-negative (TN) breast carcinomas (>40%) and BRCA1-associated TN carcinomas (>50%). These findings were confirmed in the TCGA dataset, which revealed positive associations with luminal progenitor genes (GABRP, SLC6A14, FOXC1, PROM1, and BBOX1) and strong negative correlations with ER-associated genes (ESR1, C6ORF211, GATA3, and FOXA1). Moreover, VGLL1 expression was associated with reduced overall survival. In conclusion, VGLL1 and miR-934 are mainly expressed in sporadic and BRCA1-associated TN basal-like breast carcinomas, and their coordinated expression, at least partially mediated by the direct modulation of ESR1, might be involved in the maintenance of a luminal progenitor phenotype.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining