Merck
  • Protective effect of ((4-tert-butylcyclohexylidene) methyl) (4-methoxystyryl) sulfide, a novel unsymmetrical divinyl sulfide, on an oxidative stress model induced by sodium nitroprusside in mouse brain: involvement of glutathione peroxidase activity.
Protective effect of ((4-tert-butylcyclohexylidene) methyl) (4-methoxystyryl) sulfide, a novel unsymmetrical divinyl sulfide, on an oxidative stress model induced by sodium nitroprusside in mouse brain: involvement of glutathione peroxidase activity.
The Journal of pharmacy and pharmacology (2014-08-19)
Francine R Ianiski, Catiane B Alves, Mariana M Bassaco, Claudio C Silveira, Cristiane Luchese
ZUSAMMENFASSUNG
In this study, the antioxidant action of ((4-tert-butylcyclohexylidene) methyl) (4-methoxystyryl) sulfide, a novel unsymmetrical divinyl sulfide, against oxidative damage induced by sodium nitroprusside (SNP) in brains of mice was investigated. Mice received SNP (0.335 μmol/site, intracerebroventricular) 30 min after administration of sulfide (10 mg/kg, intragastrically). After 1 h, animals were sacrificed and the brains were removed to biochemistry analysis. Thiobarbituric acid reactive species (TBARS), protein carbonyl (PC) and non-protein thiol (NPSH) levels, as well as catalase (CAT), superoxide dismutase (SOD), glutathione peroxidase (GPx), glutathione reductase (GR) and glutathione S-transferase (GST) activities were determined. SNP increased TBARS and PC levels, CAT, GPx, GR and GST activities and reduced NPSH levels. Administration of the sulfide attenuated the changes produced by SNP and increased per se GPx activity in brains of mice. Toxicological parameters revealed that this compound did not cause acute renal or hepatic damage. In conclusion, ((4-tert-butylcyclohexylidene) methyl) (4-methoxystyryl) sulfide protected against oxidative damage caused by SNP in mouse brain. GPx activity is involved, at least in part, in the cerebral antioxidant action of this compound.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
L-Glutathion, oxidiert, ≥98% (HPLC)
Sigma-Aldrich
2,4-Dinitrophenylhydrazin, reagent grade, 97%
Sigma-Aldrich
1-Chlor-2,4-dinitrobenzol, ≥99%
Sigma-Aldrich
1-Chlor-2,4-dinitrobenzol, 97%
Supelco
Glutathion, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathion, oxidiert, ≥98%, lyophilized powder
Sigma-Aldrich
L-Glutathion, oxidiert, BioXtra, ≥98%
SAFC
L-Glutathion, oxidiert
Sigma-Aldrich
L-Glutathion, oxidiert Dinatriumsalz, ≥98%, powder
Sigma-Aldrich
2-Nitro-benzoesäure, 95%, Contains 3- and 4-isomers
Glutathion, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutathion, oxidiert Dinatriumsalz, suitable for cell culture, BioReagent