Merck
  • Shorter leukocyte telomere length is independently associated with poor survival in patients with bladder cancer.
Shorter leukocyte telomere length is independently associated with poor survival in patients with bladder cancer.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2014-09-23)
Alessia Russo, Federica Modica, Simonetta Guarrera, Giovanni Fiorito, Barbara Pardini, Clara Viberti, Alessandra Allione, Rossana Critelli, Andrea Bosio, Giovanni Casetta, Giuseppina Cucchiarale, Paolo Destefanis, Paolo Gontero, Luigi Rolle, Andrea Zitella, Dario Fontana, Bruno Frea, Paolo Vineis, Carlotta Sacerdote, Giuseppe Matullo
ZUSAMMENFASSUNG
Shorter telomere length (TL) has been reported to be associated with increased risk of early death in elder individuals. Telomere shortening has been also related to chromosomal instability, which may possibly contribute to the development of several types of digestive or urogenital system cancers and smoking-related tumors. Therefore, we investigated the impact of TL on bladder cancer survival. TL was measured in leukocyte DNA from whole peripheral blood using quantitative real-time PCR in 463 patients with bladder cancer from a total 726 cases who were followed for up to 18 years. Patients with muscle-invasive tumor/any grade had shorter telomere than patients with non-muscle-invasive tumor/high-grade and with non-muscle-invasive tumor/non-high-grade (TL reference 0.7 ± 0.2; vs. respectively, 0.8 ± 0.2, P = 3.4 × 10(-2) and 0.8 ± 0.2, P = 3.6 × 10(-2)). Moreover, patients in the lowest quartiles of TL were associated with decreased survival after diagnosis (log-rank test, P = 3.9 × 10(-4)). A Cox regression adjusted by age, cancer aggressiveness, Bacillus Calmette-Guérin, radical cystectomy, radiotherapy, and chemotherapy showed an independent effect of TL on bladder cancer survival (HR, 3.9; 95% confidence interval, 1.7-9.1; P = 1.2 × 10(-3)). Our results suggest that leukocyte TL is only partly related to tumor aggressiveness and that shorter telomeres act as independent prognostic predictor of survival in patients with bladder cancer. TL information may allow to better select therapeutic approaches in patients with the same stage and grade. Blood leukocyte TL levels could provide an additional noninvasive prognostic marker to better predict survival and personalize therapies in patients with bladder cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Natriumlaurylsulfat, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri