Merck
Mechanisms of chemical cooperative carcinogenesis by epidermal Langerhans cells.
The Journal of investigative dermatology (2014-09-19)
Julia M Lewis, Christina D Bürgler, Juliet A Fraser, Haihui Liao, Kseniya Golubets, Cynthia L Kucher, Peter Y Zhao, Renata B Filler, Robert E Tigelaar, Michael Girardi
ZUSAMMENFASSUNG
Cutaneous squamous cell carcinoma (SCC) is the most prevalent invasive malignancy with metastatic potential. The epidermis is exposed to a variety of environmental DNA-damaging chemicals, principal among which are polyaromatic hydrocarbons (PAHs) ubiquitous in the environment, tobacco smoke, and broiled meats. Langerhans cells (LCs) comprise a network of dendritic cells situated adjacent to basal, suprabasal, and follicular infundibular keratinocytes that when mutated can give rise to SCC, and LC-intact mice are markedly more susceptible than LC-deficient mice to chemical carcinogenesis provoked by initiation with the model PAH, 7,12-dimethylbenz[a]anthracene (DMBA). LCs rapidly internalize and accumulate DMBA as numerous membrane-independent cytoplasmic foci. Repopulation of LC-deficient mice using fetal liver LC-precursors restores DMBA-induced tumor susceptibility. LC expression of p450 enzyme CYP1B1 is required for maximal rapid induction of DNA-damage within adjacent keratinocytes and their efficient neoplastic transformation; however, effects of tumor progression also attributable to the presence of LC were revealed as CYP1B1 independent. Thus, LCs make multifaceted contributions to cutaneous carcinogenesis, including via the handling and metabolism of chemical mutagens. Such findings suggest a cooperative carcinogenesis role for myeloid-derived cells resident within cancer susceptible epithelial tissues principally by influencing early events in malignant transformation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Ammoniumthiocyanat, ACS reagent, ≥97.5%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Supelco
Aceton, analytical standard
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Ammoniumthiocyanat, crystals
USP
Aceton, United States Pharmacopeia (USP) Reference Standard
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ammoniumthiocyanat, 99.99% trace metals basis
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%