Merck
SAS-1 is a C2 domain protein critical for centriole integrity in C. elegans.
PLoS genetics (2014-11-21)
Lukas von Tobel, Tamara Mikeladze-Dvali, Marie Delattre, Fernando R Balestra, Simon Blanchoud, Susanne Finger, Graham Knott, Thomas Müller-Reichert, Pierre Gönczy
ZUSAMMENFASSUNG
Centrioles are microtubule-based organelles important for the formation of cilia, flagella and centrosomes. Despite progress in understanding the underlying assembly mechanisms, how centriole integrity is ensured is incompletely understood, including in sperm cells, where such integrity is particularly critical. We identified C. elegans sas-1 in a genetic screen as a locus required for bipolar spindle assembly in the early embryo. Our analysis reveals that sperm-derived sas-1 mutant centrioles lose their integrity shortly after fertilization, and that a related defect occurs when maternal sas-1 function is lacking. We establish that sas-1 encodes a C2 domain containing protein that localizes to centrioles in C. elegans, and which can bind and stabilize microtubules when expressed in human cells. Moreover, we uncover that SAS-1 is related to C2CD3, a protein required for complete centriole formation in human cells and affected in a type of oral-facial-digital (OFD) syndrome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Supelco
Aceton, analytical standard
Sigma-Aldrich
Monoklonales Anti-Tubulin, acetyliert in Maus hergestellte Antikörper, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Aceton, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%