Merck
  • NADPH oxidase 4-derived H2O2 promotes aberrant retinal neovascularization via activation of VEGF receptor 2 pathway in oxygen-induced retinopathy.
NADPH oxidase 4-derived H2O2 promotes aberrant retinal neovascularization via activation of VEGF receptor 2 pathway in oxygen-induced retinopathy.
Journal of diabetes research (2015-04-14)
Jingming Li, Joshua J Wang, Sarah X Zhang
ZUSAMMENFASSUNG
NADPH oxidase 4 (Nox4) is a major isoform of NADPH oxidase in retinal endothelial cells. Our previous study suggests that upregulation of Nox4 in retinal endothelial cells contributes to retinal vascular leakage in diabetes. In the current study, we investigated the role and mechanism of Nox4 in regulation of retinal neovascularization (NV), a hallmark of proliferative diabetic retinopathy (PDR), using a mouse model of oxygen-induced retinopathy (OIR). Our results confirmed that Nox4 was expressed predominantly in retinal vasculature of mouse retina. Retinal expression of Nox4 was markedly increased in OIR, in parallel with enhanced phosphorylation of ERK. In human retinal microvascular endothelial cells (HRECs), overexpression of Nox4 by adenovirus significantly increased extracellular H2O2 generation, resulting in intensified VEGFR2 activation and exacerbated angiogenesis upon VEGF stimulation. In contrast, silencing Nox4 expression or scavenging H2O2 by polyethylene glycol- (PEG-) conjugated catalase inhibited endothelial migration, tube formation, and VEGF-induced activation of VEGFR2 signaling. Importantly, knockdown of retinal Nox4 by adenovirus-delivered siRNA significantly reduced ERK activation and attenuated retinal NV formation in OIR. Taken together, our data indicate that Nox4 promotes retinal NV formation through H2O2/VEGFR2/ERK signaling pathway. Reducing retinal Nox4 expression may represent a promising therapeutic approach for neovascular retinal diseases such as PDR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
L-Lysin -monohydrochlorid, reagent grade, ≥98% (HPLC)
Supelco
L-Lysin -monohydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysin -monohydrochlorid, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, tested according to Ph. Eur.
Lysin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysin-hydrochlorid -Lösung, 100 mM amino acid in 0.1 M HCl, analytical standard
Supelco
L-Lysin -monohydrochlorid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland