Merck
  • Characterization and storage stability of the extract of Thai mango (Mangifera indica Linn. Cultivar Chok-Anan) seed kernels.
Characterization and storage stability of the extract of Thai mango (Mangifera indica Linn. Cultivar Chok-Anan) seed kernels.
Journal of food science and technology (2014-08-13)
Pitchaon Maisuthisakul, Michael H Gordon
ZUSAMMENFASSUNG
Qualitative analysis of hydrolysable extract from mango (Mangifera indica Linn. cultivar Chok-Anan) seed kernel was performed by means of reversed phase high-performance liquid chromatography coupled to diode array detection and electrospray ionization mass spectrometry (RPHPLC-DAD-ESI-MS). The main phenolic compound was identified as methyl gallate by comparing their retention time, UV-vis absorption spectra and mass spectra with a reference standard. Quantification of phenolic compounds was performed by HPLC-DAD, which revealed that the extract contained total phenolics at a concentration of 194.1 mg GAE/g dry weight of mango seed kernel (MSK), of which 85.7% was identified as methyl gallate. In addition, the antioxidant activities of the extract and the main compound were assessed by the 2,2-diphenyl-1-picrylhydrazyl and 2,2'-azino-bis (3-ethylbenzothiazoline-6-sulfonic acid) assays, by the ferric thiocyanate method and by an assay of metal chelating activity. Tyrosinase inhibition was also investigated. Furthermore, the antioxidant capacity and the total phenolic content of MSK extract stored in a plastic (polyethylene) PE bag decreased during storage at freezing (-20 °C), refrigerated (7 °C) and room (28-32 °C) temperature for 182 days. The loss of antioxidant capacity and total phenolic content increased at higher storage temperatures for more than 182 days.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumcarbonat, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Natriumcarbonat, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchroman-2-carbonsäure, 97%
Sigma-Aldrich
Natriumcarbonat, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Natriumcarbonat, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumcarbonat, powder, 99.999% trace metals basis
Sigma-Aldrich
Natriumcarbonat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumcarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Supelco
Natriumcarbonat-Konzentrat, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Supelco
Natriumcarbonat-Konzentrat, 0.1 M Na2CO3 in water, eluent concentrate for IC
Supelco
Natriumcarbonat, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Natriumcarbonat, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Gallussäure-methylester, 98%
Sigma-Aldrich
Natriumcarbonat-12C, 99.9 atom % 12C