Merck
  • Lung transplantation from donors outside standard acceptability criteria--are they really marginal?
Lung transplantation from donors outside standard acceptability criteria--are they really marginal?
Transplant international : official journal of the European Society for Organ Transplantation (2014-07-30)
Bartlomiej Zych, Diana García Sáez, Anton Sabashnikov, Fabio De Robertis, Mohamed Amrani, Toufan Bahrami, Prashant N Mohite, Nikhil P Patil, Alexander Weymann, Aron F Popov, Anna Reed, Martin Carby, André R Simon
ZUSAMMENFASSUNG
Lung transplantation (LTx) from "extended donor criteria" donors may reduce significantly organ shortage. However, its influence on results remains unclear. In this study, we evaluate retrospectively the results of LTx from donors outside standard criteria: PaO2/FiO2 ratio < 300 mmHg, age over 55 years, and history of smoking > 20 pack-years. Two hundred and forty-eight patients underwent first time LTx in our institution between January 2007 and January 2013. Seventy-nine patients (Group I) received organs from "extended donor criteria" and 169 patients (Group II) from "standard donor criteria." Recipients' and donors' demographics, perioperative variables, and outcome were compared. Donors from Group I were significantly older [median (interquartile range)]: 52.5 (44;58) vs. 42 (28.5;48.5) years (P < 0.001) with lower PaO2/FiO2 ratio: 366 ± 116.1 455 ± 80.5 mmHg (P < 0.001), higher incidence of smoking history: 57.7% vs. 41.8% (P = 0.013), and more extensive smoking history: 24(15;30) vs. 10(3.75;14) pack-years (P < 0.001). Other parameters were comparable. Recipients' gender, diagnosis, percentage of patients operated on pump and receiving double LTx were also comparable. Recipients from Group I were significantly older: 50 (42;57) vs. 44 (29.5;53.5) years (P = 001). There were no differences observed in recipients' prevalence of primary graft dysfunction (PGD) grade 3 over first three postoperative days, duration of mechanical ventilation, intensive care and hospital length of stay, prevalence of rejection, and bronchiolitis obliterans syndrome (BOS). 90-day, 1-year, and 5-year survival (Group I vs. II) were also similar: 88.6% vs. 91.7%, 83.2% vs. 84.6%, and 59% vs. 68.2% (log rank P = 0.367). Carefully selected donor lungs from outside the standard acceptability criteria may expand existing donor pool with no detrimental effect on LTx outcome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mycophenolat-Mofetil, ≥98% (HPLC)
USP
Mycophenolat-Mofetil, United States Pharmacopeia (USP) Reference Standard
Supelco
Mycophenolat-Mofetil, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Voriconazol, VETRANAL®, analytical standard
Voriconazol, European Pharmacopoeia (EP) Reference Standard
Mycophenolat-Mofetil, European Pharmacopoeia (EP) Reference Standard
Mycophenolatmofetil für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard