Merck
  • Induction of human defensins by intestinal Caco-2 cells after interactions with opportunistic Candida species.
Induction of human defensins by intestinal Caco-2 cells after interactions with opportunistic Candida species.
Microbes and infection (2013-10-08)
Attila Gácser, Zoltán Tiszlavicz, Tibor Németh, György Seprényi, Yvette Mándi
ZUSAMMENFASSUNG
In this study we investigated the effects of Candida albicans, Candida krusei, Candida tropicalis and Candida parapsilosis on human beta-defensin 2 (HBD-2) production in Caco-2 intestinal cell line, and the production of alpha-defensins (human neutrophil peptides, HNP 1-3) in peripheral blood. Opportunistic pathogen yeasts can modulate the host immune function by inducing defensins, the natural antimicrobial peptides. Here we show that Candida spp. stimulated HBD-2 expression in and release from Caco-2 cells, with C. albicans inducing the highest levels of HBD-2. Similarly, HNP 1-3 secretion was significantly increased in whole blood after exposure to Candida yeast cells, with C. albicans producing the greatest effect. Our investigations underscore the important role of beta and alpha defensins produced by intestinal epithelial cells locally and neutrophils systemically in the antifungal defense against Candida.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Supelco
Aceton, analytical standard
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Aceton, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%