Merck
  • Synthesis, molecular modeling and NAD(P)H:quinone oxidoreductase 1 inducer activity of novel cyanoenone and enone benzenesulfonamides.
Synthesis, molecular modeling and NAD(P)H:quinone oxidoreductase 1 inducer activity of novel cyanoenone and enone benzenesulfonamides.
Journal of enzyme inhibition and medicinal chemistry (2014-01-15)
Mostafa M Ghorab, Maureen Higgins, Mansour S Alsaid, Reem K Arafa, Abdelaaty A Shahat, Albena T Dinkova-Kostova
ZUSAMMENFASSUNG
Abstract In biological systems, the Keap1/Nrf2/antioxidant response element pathway determines the ability of mammalian cells to adapt and survive conditions of oxidative, electrophilic and inflammatory stress by regulating the production of cytoprotective enzymes quinone oxidoreductase 1 (NQO1, EC 1.6.99.2) being one of them. Novel biologically active benzenesulfonamides 2, 3, 5-7, penta-2,4-dienamide 4 and chromene-2-carboxamide 8 structurally augmented with an electron-deficient Michael acceptor enone or cyanoenone functionalities were prepared. A new biological activity was conferred to these molecules, that of induction of NQO1. The potency of induction was increased by incorporation of a nitrile group adjacent to the enone and the dinitrophenyl derivative 3 was the most promising inducer. Also, molecular docking of the new compounds in the Nrf2-binding site of Keap1 was performed to assess their ability to inhibit Keap1 which biologically leads to a consequent Nrf2 accumulation and enhanced gene expression of NQO1. Docking results showed considerable interactions between the new molecules and essential binding site amino acids.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.5 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethylsulfoxid-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D