Merck
The effects of histone deacetylase inhibitors on glioblastoma-derived stem cells.
Journal of molecular neuroscience : MN (2014-05-31)
Angel A Alvarez, Melvin Field, Sergey Bushnev, Matthew S Longo, Kiminobu Sugaya
ZUSAMMENFASSUNG
Glioblastoma multiforme (GBM) is the most malignant brain tumor with limited effective treatment options. Cancer stem cells (CSCs), a subpopulation of cancer cells with stem cell properties found in GBMs, have been shown to be extremely resistant to radiation and chemotherapeutic agents and have the ability to readily reform tumors. Therefore, the development of therapeutic agents targeting CSCs is extremely important. In this study, we isolated glioblastoma-derived stem cells (GDSCs) from GBM tissue removed from patients during surgery and analyzed their gene expression using quantitative real-time PCR and immunocytochemistry. We examined the effects of histone deacetylase inhibitors trichostatin A (TSA) and valproic acid (VPA) on the proliferation and gene expression profiles of GDSCs. The GDSCs expressed significantly higher levels of both neural and embryonic stem cell markers compared to GBM cells expanded in conventional monolayer cultures. Treatment of GDSCs with histone deacetylase inhibitors, TSA and VPA, significantly reduced proliferation rates of the cells and expression of the stem cell markers, indicating differentiation of the cells. Since differentiation into GBM makes them susceptible to the conventional cancer treatments, we posit that use of histone deacetylase inhibitors may increase efficacy of the conventional cancer treatments for eliminating GDSCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Supelco
Valproinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Valproinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propylpentansäure
Valproinsäure, European Pharmacopoeia (EP) Reference Standard
Valproinsäure für die Systemeignung, European Pharmacopoeia (EP) Reference Standard