Merck
  • One pot synthesis of water-dispersible dehydroascorbic acid coated Fe3O4 nanoparticles under atmospheric air: blood cell compatibility and enhanced magnetic resonance imaging.
One pot synthesis of water-dispersible dehydroascorbic acid coated Fe3O4 nanoparticles under atmospheric air: blood cell compatibility and enhanced magnetic resonance imaging.
Journal of colloid and interface science (2014-06-24)
Hariom Gupta, P Paul, Naresh Kumar, Seema Baxi, Dipti P Das
ZUSAMMENFASSUNG
Water dispersible and biologically important molecule dehydroascorbic acid (DHA, capable to cross the blood brain barrier) coated Fe3O4 superparamagnetic nanoparticles having an average size of ∼6 nm were synthesized through one pot aqueous coprecipitation method under atmospheric air. An antioxidant ascorbic acid (AA) used in the synthesis oxidized itself to dehydroascorbic acid (DHA) to consume dissolved or available oxygen in reaction mixture which died away the oxidative impact of atmospheric air and formed DHA encapsulated the Fe3O4 nanoparticles which stabilized the Fe3O4 nanoparticles and significantly enhanced their colloidal solubility in water. Fe3O4 phase, superparamagnetic property, DHA coating and stable colloidal solubility in water were confirmed by means of XPS, VSM, IR and zeta potential analysis respectively. T1, T2 and T2(∗) weighted magnetic resonance imaging (MRI) and corresponding relaxivity (r1=0.416, r2=50.28 and r2(∗)=123.65 mM(-1) and r2/r1=120.86, r2(∗)r1=297.23) of colloidally dispersed DHA-coated nanoparticle water phantom revealed a strong contrast enhancement in T2 and T2(∗) weighted images. The compatibility of DHA-coated Fe3O4 nanoparticles toward human blood cells was examined by means of cell counting and cell morphological analysis with the use of optical microscope and scanning electron microscope imaging.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Kaliumbromid, ≥99% trace metals basis
Sigma-Aldrich
Eisen(II)-sulfat Heptahydrat, ReagentPlus®, ≥99%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Eisen(II)-sulfat Heptahydrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Kaliumbromid, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Osmiumtetroxid, ReagentPlus®, 99.8%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Osmiumtetroxid -Lösung, 4 wt. % in H2O
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Eisen(II)-sulfat Heptahydrat, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Supelco
Natronlauge, 49-51% in water, eluent for IC
Sigma-Aldrich
Kaliumbromid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Osmiumtetroxid -Lösung, 2.5 wt. % in tert-butanol
Supelco
Natronlauge-Konzentrat, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Osmiumtetroxid, Sealed ampule.
Sigma-Aldrich
Osmiumtetroxid -Lösung, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets