Merck
  • A method to prevent protein delocalization in imaging mass spectrometry of non-adherent tissues: application to small vertebrate lens imaging.
A method to prevent protein delocalization in imaging mass spectrometry of non-adherent tissues: application to small vertebrate lens imaging.
Analytical and bioanalytical chemistry (2015-02-11)
David M G Anderson, Kyle A Floyd, Stephen Barnes, Judy M Clark, John I Clark, Hassane Mchaourab, Kevin L Schey
ZUSAMMENFASSUNG
MALDI imaging requires careful sample preparation to obtain reliable, high-quality images of small molecules, peptides, lipids, and proteins across tissue sections. Poor crystal formation, delocalization of analytes, and inadequate tissue adherence can affect the quality, reliability, and spatial resolution of MALDI images. We report a comparison of tissue mounting and washing methods that resulted in an optimized method using conductive carbon substrates that avoids thaw mounting or washing steps, minimizes protein delocalization, and prevents tissue detachment from the target surface. Application of this method to image ocular lens proteins of small vertebrate eyes demonstrates the improved methodology for imaging abundant crystallin protein products. This method was demonstrated for tissue sections from rat, mouse, and zebrafish lenses resulting in good-quality MALDI images with little to no delocalization. The images indicate, for the first time in mouse and zebrafish, discrete localization of crystallin protein degradation products resulting in concentric rings of distinct protein contents that may be responsible for the refractive index gradient of vertebrate lenses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenylephrin-Hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sinapinsäure, ≥98%, powder
Sigma-Aldrich
(R)-(−)-Phenylephrin -hydrochlorid, powder
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
USP
Phenylephrin -hydrochlorid, United States Pharmacopeia (USP) Reference Standard
Supelco
Sinapinsäure, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Supelco
Sinapinsäure, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Tropicamid, solid
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Phenylephrin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Phenylephrinhydrochlorid für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Supelco
trans-Sinapinsäure, analytical standard
Sigma-Aldrich
(R)-(−)-Phenylephrin -hydrochlorid, analytical standard
Tropicamid, European Pharmacopoeia (EP) Reference Standard