Merck
  • A label-free amperometric immunosensor for detection of zearalenone based on trimetallic Au-core/AgPt-shell nanorattles and mesoporous carbon.
A label-free amperometric immunosensor for detection of zearalenone based on trimetallic Au-core/AgPt-shell nanorattles and mesoporous carbon.
Analytica chimica acta (2014-09-30)
Lei Liu, Yingjun Chao, Wei Cao, Yulan Wang, Chuannan Luo, Xuehui Pang, Dawei Fan, Qin Wei
ZUSAMMENFASSUNG
A novel label-free amperometric immunosensor is proposed for the ultrasensitive detection of zearalenone (ZEN) based on mesoporous carbon (MC) and trimetallic nanorattles (core/shell particles with movable cores encapsulated in the shells). The nanorattles are composed of special Au-core and imperfect AgPt-shell structure (Au@AgPt). The Au@AgPt nanorattles are loaded onto the MC by physical adsorption. The structure of the Au@AgPt nanorattles was characterized by using scanning electron microscopy (SEM) and transmission electron microscopy (TEM). Energy dispersive X-ray spectroscopy (EDS) confirmed the composition of the synthesized nanorattles. Compared with monometallic and bimetallic nanoparticles (NPs), Au@AgPt nanorattles show a higher electron transfer rate due to the synergistic effect of the Au, Ag and Pt NPs. MC further improves the sensitivity of the immunosensor because of its extraordinarily large specific surface area, suitable pore arrangement and outstanding conductivity. The large specific surface area of MC and MC@Au@AgPt were characterized by the BET method. ZEN antibodies are immobilized onto the nanorattles via Ag-NH2 bonds and Pt-NH2 bonds. Cyclic voltammetry and square wave voltammetry were used to characterize the recognizability of ZEN. Under optimum experimental conditions, the proposed immunosensor exhibited a low detection limit (1.7 pg mL(-1)), a wide linear range (from 0.005 to 15 ng mL(-1)) as well as good stability, reproducibility and selectivity. The sensor can be used in clinical analysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, Molecular Biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥96.0% (AT)
USP
Cetrimoniumbromid, United States Pharmacopeia (USP) Reference Standard
Supelco
Hexadecyltrimethylammoniumbromid, analytical standard
Supelco
Hexadecyltrimethylammoniumbromid, suitable for ion pair chromatography, LiChropur