Merck
  • Evaluation of a ⁶⁴Cu‑labeled 1,4,7‑triazacyclononane, 1‑glutaric acid‑4,7 acetic acid (NODAGA)‑galactose‑bombesin analogue as a PET imaging probe in a gastrin‑releasing peptide receptor‑expressing prostate cancer xenograft model.
Evaluation of a ⁶⁴Cu‑labeled 1,4,7‑triazacyclononane, 1‑glutaric acid‑4,7 acetic acid (NODAGA)‑galactose‑bombesin analogue as a PET imaging probe in a gastrin‑releasing peptide receptor‑expressing prostate cancer xenograft model.
International journal of oncology (2015-01-15)
Min Hwan Kim, Ji Ae Park, Sang-Keun Woo, Kyo Chul Lee, Gwang Il An, Byoung Soo Kim, Kwang Il Kim, Tae Sup Lee, Chan Wha Kim, Kyeong Min Kim, Joo Hyun Kang, Yong Jin Lee
ZUSAMMENFASSUNG
Gastrin‑releasing peptide receptor (GRPR) is overexpressed by a variety of human tumors and in particular, identified to be upregulated in prostate cancers. The current study aimed to develop clinically translatable BBN analogue‑based radioligands for positron emission tomography (PET) of GRPR‑positive tumors. We developed radiolabeled BBN analogues and modified radiolabeled galacto‑BBN analogues and then investigated their tumor‑targeting efficacy in vivo. The chelator 1,4,7‑triazacyclononane, 1‑glutaric acid‑4,7 acetic acid (NODAGA) was used to radiolabel the peptides with 64Cu. The peptides were evaluated by measuring cell‑based receptor‑binding affinities. Biodistribution experiments and small animal imaging using PET were performed in nude mice bearing subcutaneous PC3 human prostate cancer xenografts. The conjugates were radiolabeled with yields >99%. The stability assay showed that [64Cu]NODAGA‑BBN and [64Cu]NODAGA‑galacto‑BBN remained stable in both human and mouse serum for 1 h at 37˚C. PET images of PC3 tumor‑bearing nude mice were acquired at 1, 3, 24, 48 and 72 h after injection. [64Cu]NODAGA‑galacto‑BBN showed retention in tumors for 72 h, low liver uptake, and rapid renal clearance. PET imaging results were also confirmed by biodistrubution 1 and 3 h after injection. [64Cu]NODAGA‑BBN and [64Cu]NODAGA‑galacto‑BBN are promising new PET probes for GRPR‑positive prostate cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Natriumacetat, ACS reagent, ≥99.0%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Natriumacetat, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Sigma-Aldrich
Natriumacetat, >99%, FG
Sigma-Aldrich
Trizma® Base, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® Base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® Base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Natriumacetat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Tantal(V)-ethoxid, 99.98% trace metals basis
Sigma-Aldrich
Natriumacetat, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumacetat -Lösung, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Natriumacetat, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Natriumacetat, meets USP testing specifications, anhydrous
Supelco
Tromethamin, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Trizma® Base, 99.7% (T), puriss. p.a.
Supelco
Trizma® Base, reference material for titrimetry, certified by BAM, >99.5%
USP
Tromethamin, United States Pharmacopeia (USP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Trometamol, European Pharmacopoeia (EP) Reference Standard
SAFC
Tromethamin
Sigma-Aldrich
Natriumacetat, BioXtra, ≥99.0%