Merck
  • Loss of p53 in stromal fibroblasts enhances tumor cell proliferation through nitric-oxide-mediated cyclooxygenase 2 activation.
Loss of p53 in stromal fibroblasts enhances tumor cell proliferation through nitric-oxide-mediated cyclooxygenase 2 activation.
Free radical research (2014-12-17)
S Wada, Y Matsushita, H Tazawa, W Aoi, Y Naito, A Higashi, H Ohshima, T Yoshikawa
ZUSAMMENFASSUNG
Overexpression of cyclooxygenase 2 (COX-2) by stromal fibroblasts plays a critical role in the early stage of carcinogenesis. COX-2 expression is thought to be positively or negatively regulated by inflammatory chemical mediators or tumor suppressors. In this study, the contributions of inducible nitric oxide synthase (iNOS) and p53 to COX-2 expression were examined using mouse embryonic fibroblasts (MEFs) from wild-type, p53-deficient, iNOS-deficient, and p53/iNOS-deficient mice. These MEFs were treated with 1 μg/mL of lipopolysaccharide and 100 IU/mL of interferon gamma for up to 72 h. iNOS and COX-2 expression were analyzed by Western blotting. iNOS was induced earlier (16 h) in p53-deficient MEFs than in wild-type MEFs (48 h). Elevated expression of COX-2 was sustained for a longer duration in the p53-deficient MEFs. In contrast, COX-2 expression was reduced earlier in the iNOS-deficient MEFs. Addition of an exogenous NO donor (0.8 mM of S-nitroso-l-glutathione) to the iNOS-deficient MEFs augmented COX-2 expression. Co-culture with stimulated p53-deficient MEFs promoted cell proliferation of mouse rectal polyploid carcinoma CMT93 cells, but treatment with a COX-2-specific inhibitor counteracted this effect. These results suggest that loss of function of the p53 gene in stromal fibroblasts enhances COX-2 expression by enhancing iNOS expression and the resultant production of NO, contributing to the promotion of tumor growth.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
N-(1-Naphthyl)ethylendiamin -dihydrochlorid, ACS reagent, >98%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sulfanilamid, ≥98%
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
N-(1-Naphthyl)ethylendiamin -dihydrochlorid, ≥98%
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Chlorwasserstoff – Methanol -Lösung, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Supelco
Sulfanilamid Schmelzpunktstandard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in acetic acid
Sigma-Aldrich
Sulfanilamid, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
S-Nitrosoglutathion, ≥97%
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Supelco
N-(1-Naphthyl)ethylendiamin -dihydrochlorid, ACS reagent, ≥98%
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
USP
Sulfanilamid, United States Pharmacopeia (USP) Reference Standard