Merck
  • Uncarboxylated osteocalcin stimulates 25-hydroxy vitamin D production in Leydig cell line through a GPRC6a-dependent pathway.
Uncarboxylated osteocalcin stimulates 25-hydroxy vitamin D production in Leydig cell line through a GPRC6a-dependent pathway.
Endocrinology (2014-08-06)
Luca De Toni, Vincenzo De Filippis, Simone Tescari, Marco Ferigo, Alberto Ferlin, Valentina Scattolini, Angelo Avogaro, Roberto Vettor, Carlo Foresta
ZUSAMMENFASSUNG
Recent studies disclosed a cross talk between testis and bone. By the action of LH, Leydig cells are able to modulate bone metabolism through testosterone and insulin-like factor 3. Moreover, LH modulates the Leydig expression of CYP2R1, the key enzyme involved in vitamin D (Vit D) 25-hydroxylation. However, pathways regulating CYP2R1 expression have been poorly investigated. The cross talk from the bone to the testis of the vitamin D 25-hydroxylase CYP2R1 involves osteocalcin (OC), which is produced by the osteoblasts and stimulates the production of testosterone by the Leydig cells through its putative receptor GPRC6A, a cation-sensing G-protein-coupled receptor. The aim of this study was to investigate the possible action of OC on CYP2R1 expression and 25-hydroxy Vit D (25-OH Vit D) production in a mouse Leydig cell line (MA-10). After confirmation of the expression of GPRC6A by MA-10, we found that stimulation with either human chorionic gonadotropin or uncarboxylated-OC (ucOC) increases CYP2R1 protein expression in a dose-dependent manner and, in turn, increases the release of 25-OH Vit D in culture medium. This effect was abolished by receptor blockade with, respectively, anti-LH receptor and anti-GPRC6A antibodies. Moreover, both agonists converged to phosphorylation of Erk1/2 by a likely differential action on second messengers. Human chorionic gonadotropin induced slow "tonic" increase of intercellular calcium and accumulation of cAMP, whereas ucOC mainly induced phasic increase of cell calcium. Supporting these findings, we found that serum ucOC positively correlated with 25-OH Vit D levels in 40 overweight male patients and 21 controls. Altogether, our results suggest that OC contributes with LH to 25-OH Vit D production by Leydig cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Zyklisches Adenosin-3′,5′-Monophosphat, ≥98.5% (HPLC), powder
Supelco
Cholecalciferol (D3), analytical standard
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Supelco
(±)-Sulfinpyrazon, analytical standard
Sulfinpyrazon, European Pharmacopoeia (EP) Reference Standard
Sulfinpyrazon für die Systemeignung, European Pharmacopoeia (EP) Reference Standard