Merck
  • Radiofrequency ablation for the treatment of recurrent bone and soft-tissue sarcomas in non-surgical candidates.
Radiofrequency ablation for the treatment of recurrent bone and soft-tissue sarcomas in non-surgical candidates.
International journal of clinical oncology (2013-11-30)
Koichiro Yamakado, Akihiko Matsumine, Tomoki Nakamura, Atsuhiro Nakatsuka, Haruyuki Takaki, Takao Matsubara, Kunihiro Asanuma, Akihiro Sudo, Yoshiki Sugimura, Hajime Sakuma
ZUSAMMENFASSUNG
The aim of this study was to evaluate the safety and clinical utility of radiofrequency ablation (RFA) retrospectively in non-surgical candidates with recurrent bone and soft-tissue sarcomas. Percutaneous RFA was used in 52 patients (21 female, 31 male; mean age, 52.2 ± 21.1 years; range 10-87 years) with recurrent bone and soft-tissue sarcomas. The number of tumors was 3 or fewer in 23 patients (44.2 %) and 4 or more in the others, with a mean maximum tumor diameter of 3.0 ± 3.7 cm (range 0.5-18 cm). Safety, tumor control, and prognosis were evaluated. All tumors were ablated after RFA in 21 patients (40.4 %, 21/52), although 14 experienced re-recurrence. Tumors were controlled in 8 cases of recurrence by repeat RFA (n = 7) and surgical intervention (n = 1). Therefore, 15 patients (28.8 %, 15/52) were tumor-free at the end of follow-up (mean follow-up 25.5 ± 24.2 months; range 3.9-117 months). Residual tumors were found after RFA in the other 31 patients (59.6 %, 31/52). Overall survival rates were 73.4 % (95 % CI 61.0-85.9 %) at 1 year, 39.3 % (95 % CI 23.6-54.9 %) at 3 years, and 34.3 % (95 % CI 18.0-50.7 %) at 5 years in all patients. Recurrence-free interval (p = 0.040), tumor number (p = 0.0094), and complete tumor ablation (p < 0.0001) were detected as significant prognostic factors in univariate analysis. The latter two factors were significant in multivariate analysis. The rate of major complications was 0.9 %. RFA is a safe and useful therapeutic option for treatment of recurrent bone and soft-tissue sarcomas. Prognostic factors found in this study will help to identify those patients who would benefit from RFA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
USP
Cefazolin, United States Pharmacopeia (USP) Reference Standard
Cefazolin, European Pharmacopoeia (EP) Reference Standard