Merck
  • Plasma HSP70 levels correlate with health risk factors and insulin resistance in African American subjects.
Plasma HSP70 levels correlate with health risk factors and insulin resistance in African American subjects.
Experimental and clinical endocrinology & diabetes : official journal, German Society of Endocrinology [and] German Diabetes Association (2014-05-21)
A Islam, S H Hait, B Andrews-Shigaki, S Carus, P A Deuster
ZUSAMMENFASSUNG
The family of heat shock proteins works at the cellular level to protect cells against many chronically and acutely induced stressful conditions. The induction of certain family members, such as HSP70 and HSP90 via a heat shock factor 1 activated pathway, may have broad therapeutic benefits in the treatment of various types of tissue trauma and metabolic diseases. HSPs with their ability to improve cytoprotective functions and enhance tissue regeneration, offer a new potential in treating conditions such as diabetes and obesity. We hypothesized that the plasma levels of HSP70 and/or HSP90 would be significantly lower in obese than non-obese African American men and women. The objective of the study was to provide an analysis of plasma HSP70 and HSP90 concentrations of African American subjects grouped according to widely accepted health risk indices and insulin resistance. Comparisons of HSP70 and HSP90 concentrations by BMI, percent body fat, waist circumference, insulin resistance, plasma cortisol levels and gender were conducted. HSP70 concentration inversely correlated with BMI, percentage body fat, waist circumference, and insulin resistance. No significant correlation was observed for HSP90 concentration with the aforementioned indices. Our results show that high risk health conditions, such as obesity and type-2 diabetes, may be associated with compromised expression of specific heat shock proteins such as HSP70.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Supelco
Hydrocortison, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrocortison, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortison, meets USP testing specifications
Hydrocortison für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Hydrocortison, European Pharmacopoeia (EP) Reference Standard
Hydrocortison, British Pharmacopoeia (BP) Assay Standard