Merck
  • Heat shock protein-27 delays acute rejection after cardiac transplantation: an experimental model.
Heat shock protein-27 delays acute rejection after cardiac transplantation: an experimental model.
Transplantation (2014-06-01)
Borggia Seemampillai, Renee Germack, Leanne E Felkin, Ann McCormack, Marlene L Rose
ZUSAMMENFASSUNG
Rejection is the major obstacle to survival after cardiac transplantation. We investigated whether overexpression of heat shock protein (Hsp)-27 in mouse hearts protects against acute rejection and the mechanisms of such protection. Hearts from B10.A mice overexpressing human Hsp-27 (Hsp-27tg), or Hsp-27-negative hearts from littermate controls (LCs) were transplanted into allogeneic C57BL/6 mice. The immune response to B10.A hearts was investigated using quantitative polymerase chain reaction for CD3+, CD4+, CD8+ T cells, and CD14+ monocytes and cytokines (interferon-γ, interleukin [IL]-2, tumor necrosis factor-α, IL-1β, IL-4, IL-5, IL-10, transforming growth factor-β) in allografts at days 2, 5, and 12 after transplantation. The effect of Hsp-27 on ischemia-induced caspase activation and immune activation was investigated. Survival of Hsp-27tg hearts (35±10.37 days, n=10) was significantly prolonged compared with LCs (13.6±3.06 days, n=10, P=0.0004). Hsp-27tg hearts expressed significantly more messenger RNA (mRNA) markers of CD14+ monocytes at day 2 and less mRNA markers of CD3+ and CD8+T cells at day 5 compared with LCs. There was more IL-4 mRNA in Hsp-27tg hearts at day 2 and less interferon-γ mRNA at day 5 compared with LCs. Heat shock protein-27tg hearts subjected to ischemia or to 24 hr ischemia-reperfusion injury demonstrated significantly less apoptosis and activation of caspases 3, 9, and 1 than LCs. T cells removed from C57BL/6 recipients of Hsp-27tg hearts produced a vigorous memory response to B10.A antigens, suggesting immune activation was not inhibited by Hsp-27. Heat shock protein-27 delays allograft rejection, by inhibiting tissue damage, through probably an antiapoptotic pathway. It may also promote an anti-inflammatory subset of monocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, tested according to Ph. Eur.
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-7-amido-4-methylcoumarin, ≥97% (HPLC), powder