Merck
  • Differential effects of short- and long-term antipsychotic treatment on the expression of neuregulin-1 and ErbB4 receptors in the rat brain.
Differential effects of short- and long-term antipsychotic treatment on the expression of neuregulin-1 and ErbB4 receptors in the rat brain.
Psychiatry research (2015-01-13)
Chao Deng, Bo Pan, Chang-Hua Hu, Mei Han, Xu-Feng Huang
ZUSAMMENFASSUNG
Neuregulin-1 (NRG1) and ErbB4 genes have been identified as candidate genes for schizophrenia. Post-mortem studies indicated that NRG1-ErbB4 signalling is impaired in schizophrenia subjects. This study investigated whether short- or long-term antipsychotic treatment has different effects on the expression of NRG1 and ErbB4 receptors. Female Sprague-Dawley rats were treated orally with either aripiprazole (0.75 mg/kg), haloperidol (0.1 mg/kg), olanzapine (0.5 mg/kg), or vehicle, 3 times/day for 1 or 12 weeks. Western blotting was performed to examine the expression of NRG1 isoforms (135 kDa, 70 kDa and 40 kDa) and ErbB4 receptors. Both 1-week haloperidol and olanzapine treatment increased NRG1-70kDa expression in the hippocampus; haloperidol also up-regulated ErbB4 levels in the prefrontal cortex (PFC). In the 12-week group, aripiprazole decreased the expression of all three NRG1 isoforms and ErbB4 receptors in the PFC, NRG1-70 kDa and -40 kDa in the cingulate cortex (Cg), and NRG1-135 kDa, -70 kDa and ErbB4 receptors in the hippocampus; haloperidol reduced NRG1-135 kDa in the PFC, NRG1-40 kDa in all three brain regions, and ErbB4 receptor levels in the PFC and hippocampus; NRG1-40 kDa in the PFC and Cg was also down-regulated by olanzapine. These results suggest that the time-dependent and region-specific effects of antipsychotics on NRG1-ErbB4 signalling may contribute to the efficacy of antipsychotics to treat schizophrenia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Glycin
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
Haloperidol, powder
Sigma-Aldrich
Glycin, 99%, FCC
USP
Glycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Aripiprazol, ≥98% (HPLC)
Sigma-Aldrich
Olanzapin, ≥98% (HPLC)
USP
Olanzapin, United States Pharmacopeia (USP) Reference Standard