Merck
Expression pattern analysis of odorant-binding proteins in the pea aphid Acyrthosiphon pisum.
Insect science (2014-03-05)
Filomena De Biasio, Lea Riviello, Daniele Bruno, Annalisa Grimaldi, Terenzio Congiu, Yu Feng Sun, Patrizia Falabella
ZUSAMMENFASSUNG
Odorant-binding proteins (OBPs) are soluble proteins mediating chemoreception in insects. In previous research, we investigated the molecular mechanisms adopted by aphids to detect the alarm pheromone (E)-β-farnesene and we found that the recognition of this and structurally related molecules is mediated by OBP3 and OBP7. Here, we show the differential expression patterns of 5 selected OBPs (OBP1, OBP3, OBP6, OBP7, OBP8) obtained performing quantitative RT-PCR and immunolocalization experiments in different body parts of adults and in the 5 developmental instars, including winged and unwinged morphs, of the pea aphid Acyrthosiphon pisum. The results provide an overall picture that allows us to speculate on the relationship between the differential expression of OBPs and their putative function. The expression of OBP3, OBP6, and OBP7 in the antennal sensilla suggests a chemosensory function for these proteins, whereas the constant expression level of OBP8 in all instars could suggest a conserved role. Moreover, OBP1 and OBP3 are also expressed in nonsensory organs. A light and scanning electron microscopy study of sensilla on different body parts of aphid, in particular antennae, legs, mouthparts, and cornicles-cauda, completes this research providing a guide to facilitate the mapping of OBP expression profiles.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard