Merck
Transport mechanisms of a novel antileukemic and antiviral compound 9-norbornyl-6-chloropurine.
Journal of enzyme inhibition and medicinal chemistry (2014-04-01)
Pavla Plačková, Hubert Hřebabecký, Michal Šála, Radim Nencka, Tomáš Elbert, Helena Mertlíková-Kaiserová
ZUSAMMENFASSUNG
6-Chloropurines substituted at the position 9 with variously modified bicyclic skeletons represent promising antiviral and anticancer agents. This work aimed to investigate the transport mechanisms of 9-[(1R*,2R*,4S*)-bicyclo[2.2.1]hept-2-yl]-6-chloro-9H-purine (9-norbornyl-6-chloropurine, NCP) and their relationship to the metabolism and biological activity of the compound. Transport experiments were conducted in CCRF-CEM cells using radiolabeled compound ([(3)H]NCP). The pattern of the intracellular uptake of [(3)H]NCP in CCRF-CEM cells pointed to a combination of passive and facilitated diffusion as prevailing transport mechanisms. NCP intracellular metabolism was found to enhance its uptake by modifying NCP concentration gradient. The transport kinetics reached steady state under the conditions of MRP and MDR proteins blockade, indicating that NCP is a substrate for these efflux pumps. Their inhibition also increased the cytotoxicity of NCP. Our findings suggest that the novel nucleoside analog NCP has potential to become a new orally available antileukemic agent due to its rapid membrane permeation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
N-Methyl-D-Glucamin, 99.0-100.5% (titration)
Sigma-Aldrich
Cytochalasin D aus Zygosporium masonii, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Phenylessigsäure, 99%
Sigma-Aldrich
Cytochalasin D aus Zygosporium masonii, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Sigma-Aldrich
N-Methyl-D-Glucamin, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
Phenylessigsäure, ≥99%, FCC, FG
Sigma-Aldrich
Dipyridamol, ≥98% (HPLC)
Sigma-Aldrich
Ethacrynsäure, ≥97% (HPLC)
Sigma-Aldrich
Meglumin, 99.0-100.5% dry basis, meets USP testing specifications
Supelco
Meglumin, Pharmaceutical Secondary Standard; Certified Reference Material
Meglumin, European Pharmacopoeia (EP) Reference Standard
Dipyridamol für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Dipyridamol, European Pharmacopoeia (EP) Reference Standard
Ethacrynsäure, European Pharmacopoeia (EP) Reference Standard
Etacrynsäure für die Systemeignung, European Pharmacopoeia (EP) Reference Standard