Merck
  • Photocatalytic degradation of organic dyes by La³⁺/Ce³⁺-H₃PW₁₂O₄₀ under different light irradiation.
Photocatalytic degradation of organic dyes by La³⁺/Ce³⁺-H₃PW₁₂O₄₀ under different light irradiation.
Dalton transactions (Cambridge, England : 2003) (2014-05-09)
Taohai Li, Quanguo Li, Jing Yan, Feng Li
ZUSAMMENFASSUNG
New photocatalysts (La(3+)-H3PW12O40 and Ce(3+)-H3PW12O40) were prepared, and the degradation activity of these products was evaluated. These photocatalysts were synthesized by the hydrothermal method assisted by PEG-1000 from the reactions of LaCl3 or CeCl3 and phosphotungstic acid. The prepared catalysts were characterized by X-ray diffraction (XRD), Fourier transform infrared spectroscopy (FT-IR), scanning electron microscopy (SEM), ultraviolet-visible diffuse reflection spectroscopy (UV-vis) and thermal gravimetric analysis (TGA). The catalytic activity of La(3+)-H3PW12O40 and Ce(3+)-H3PW12O40 was evaluated by adopting methyl orange (MO) and Rhodamine B (RhB) as model dyes, and the reaction was carried out under UV- and visible light irradiation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rhodamin B, ≥95% (HPLC)
Sigma-Aldrich
Rhodamin B, suitable for fluorescence
Sigma-Aldrich
Methylorange, ACS reagent, Dye content 85 %
Sigma-Aldrich
Methylorange, suitable for microscopy
Supelco
Rhodamin B, analytical standard
Supelco
Rhodamin B -Lösung, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
Rhodamin B, Dye content 90 %