Merck
  • Characterization of a novel cytochrome P450 from Amycolatopsis sp. CGMCC1149 for hydroxylation of lovastatin.
Characterization of a novel cytochrome P450 from Amycolatopsis sp. CGMCC1149 for hydroxylation of lovastatin.
Biotechnology and applied biochemistry (2014-05-23)
Hong Zong, Bin Zhuge, Xinyao Lu, Xiaoyu Huo, Huiying Fang, Jian Song, Jin Sun
ZUSAMMENFASSUNG
Wuxistatin, a novel and potent statin, is converted from lovastatin by Amycolatopsis sp. CGMCC1149. In the bioconversion, lovastatin is firstly hydroxylated to 3-hydroxymethyl lovastatin (product I) by a hydroxylase. In the current study, a novel hydroxylase gene p450lov was isolated from Amycolatopsis sp. CGMCC1149 by degenerate PCR and self-formed adaptor PCR and expressed in Escherichia coli. The gene encodes a 403-amino-acid protein with a molecular weight of 44.8 kDa and was designated as a new member of cytochrome P450 (CYP) 105 family, CYP105A44. Meanwhile, a lovastatin catalytic in vitro system was established, and an optimal hydroxylation reaction system contained 30 µM lovastatin, 600 µM NADH, 120 µM ferredoxin, 0.04 U ferredoxin-nicotinamide adenine dinucleotide phosphate reductase, and 100 µM CYP105A44 in a final volume of 200 µL Tris HCl buffer (50 mM, pH 7.4). These would be helpful for further studies on the hydroxylation of statins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Supelco
Lovastatin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
USP
Lovastatin, United States Pharmacopeia (USP) Reference Standard
Lovastatin, European Pharmacopoeia (EP) Reference Standard