Merck
  • Mouse aldehyde dehydrogenase ALDH3B2 is localized to lipid droplets via two C-terminal tryptophan residues and lipid modification.
Mouse aldehyde dehydrogenase ALDH3B2 is localized to lipid droplets via two C-terminal tryptophan residues and lipid modification.
The Biochemical journal (2014-10-07)
Takuya Kitamura, Shuyu Takagi, Tatsuro Naganuma, Akio Kihara
ZUSAMMENFASSUNG
Aldehyde dehydrogenases (ALDHs) catalyse the conversion of toxic aldehydes into non-toxic carboxylic acids. Of the 21 ALDHs in mice, it is the ALDH3 family members (ALDH3A1, ALDH3A2, ALDH3B1, ALDH3B2 and ALDH3B3) that are responsible for the removal of lipid-derived aldehydes. However, ALDH3B2 and ALDH3B3 have yet to be characterized. In the present study, we examined the enzyme activity, tissue distribution and subcellular localization of ALDH3B2 and ALDH3B3. Both were found to exhibit broad substrate preferences from medium- to long-chain aldehydes, resembling ALDH3A2 and ALDH3B1. Although ALDH3B2 and ALDH3B3 share extremely high sequence similarity, their localizations differ, with ALDH3B2 found in lipid droplets and ALDH3B3 localized to the plasma membrane. Both were modified by prenylation at their C-termini; this modification greatly influenced their membrane localization and enzymatic activity towards hexadecanal. We found that their C-terminal regions, particularly the two tryptophan residues (Trp462 and Trp469) of ALDH3B2 and the two arginine residues (Arg462 and Arg463) of ALDH3B3, were important for the determination of their specific localization. Abnormal quantity and perhaps quality of lipid droplets are implicated in several metabolic diseases. We speculate that ALDH3B2 acts to remove lipid-derived aldehydes in lipid droplets generated via oxidative stress as a quality control mechanism.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)