Merck
  • Extracellular ATP induces intracellular alpha-synuclein accumulation via P2X1 receptor-mediated lysosomal dysfunction.
Extracellular ATP induces intracellular alpha-synuclein accumulation via P2X1 receptor-mediated lysosomal dysfunction.
Neurobiology of aging (2014-12-07)
Ming Gan, Simon Moussaud, Peizhou Jiang, Pamela J McLean
ZUSAMMENFASSUNG
The pathologic hallmark of Parkinson's disease (PD) is the accumulation of alpha-synuclein (αsyn) in susceptible neurons in the form of Lewy bodies and Lewy neurites. The etiology of PD remains unclear. Because brain injury has been suggested to facilitate αsyn aggregation, we investigated whether cellular breakdown products from damaged cells can act on neighboring healthy cells and cause intracellular αsyn accumulation and/or aggregation. Using 2 neuronal cell models, we found that extracellular adenosine triphosphate (ATP) induced a significant increase in intracellular αsyn levels between 24 and 48 hours after treatment. Further investigation revealed that the observed αsyn accumulation is a result of lysosome dysfunction caused by extracellular ATP-induced elevation of lysosomal pH. Interestingly, P2X1 receptor appears to mediate the cells' response to extracellular ATP. Although Ca(2+) influx via P2X1 receptor is necessary for αsyn accumulation, Ca(2+) influx per se is not sufficient for increased αsyn accumulation. These findings provide new insight into our knowledge of the role of P2X receptors in PD pathogenesis and may be helpful in identifying new therapeutic targets for PD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fibronektin-Rinderplasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
N6,2′-O-Dibutyryladenosin 3′,5′-zyklisches Monophosphat Natriumsalz Monohydrat, ≥96% (HPLC), powder
Sigma-Aldrich
Actidion, from microbial, ≥94% (TLC)
Sigma-Aldrich
Poly-L-ornithin -hydrobromid, mol wt 30,000-70,000
Sigma-Aldrich
Adenosin-5′-diphosphat Natriumsalz, bacterial, ≥95% (HPLC)
Sigma-Aldrich
Adenosin-5′-triphosphat Dinatriumsalz Hydrat, BioXtra, ≥99% (HPLC), from microbial
Sigma-Aldrich
Kalzium-Ionophor A23187, ≥98% (TLC), powder
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, ascites fluid
Sigma-Aldrich
Adenosin-5′-monophosphat Natriumsalz, from yeast, ≥99%
Sigma-Aldrich
Bafilomycin A1, Streptomyces griseus