Merck
  • Development of a multianalyte method based on micro-matrix-solid-phase dispersion for the analysis of fragrance allergens and preservatives in personal care products.
Development of a multianalyte method based on micro-matrix-solid-phase dispersion for the analysis of fragrance allergens and preservatives in personal care products.
Journal of chromatography. A (2014-04-29)
Maria Celeiro, Eugenia Guerra, J Pablo Lamas, Marta Lores, Carmen Garcia-Jares, Maria Llompart
ZUSAMMENFASSUNG
An effective, simple and low cost sample preparation method based on matrix solid-phase dispersion (MSPD) followed by gas chromatography-mass spectrometry (GC-MS) or gas chromatography-triple quadrupole-mass spectrometry (GC-MS/MS) has been developed for the rapid simultaneous determination of 38 cosmetic ingredients, 25 fragrance allergens and 13 preservatives. All target substances are frequently used in cosmetics and personal care products and they are subjected to use restrictions or labeling requirements according to the EU Cosmetic Directive. The extraction procedure was optimized on real non-spiked rinse-off and leave-on cosmetic products by means of experimental designs. The final miniaturized process required the use of only 0.1g of sample and 1 mL of organic solvent, obtaining a final extract ready for analysis. The micro-MSPD method was validated showing satisfactory performance by GC-MS and GC-MS/MS analysis. The use of GC coupled to triple quadrupole mass detection allowed to reach very low detection limits (low ng g(-1)) improving, at the same time, method selectivity. In an attempt to improve the chromatographic analysis of preservatives, the inclusion of a derivatization step was also assessed. The proposed method was applied to a broad range of cosmetics and personal care products (shampoos, body milk, moisturizing milk, toothpaste, hand creams, gloss lipstick, sunblock, deodorants and liquid soaps among others), demonstrating the extended use of these substances. The concentration levels were ranging from the sub parts per million to the parts per mill. The number of target fragrance allergens per samples was quite high (up to 16). Several fragrances (linalool, farnesol, hexylcinnamal, and benzyl benzoate) have been detected at levels >0.1% (1,000 μg g(-1)). As regards preservatives, phenoxyethanol was the most frequently found additive reaching quite high concentration (>1,500 μg g(-1)) in five cosmetic products. BHT was detected in eight samples, in two of them (a baby care product and a lipstick) at high concentrations (>1,000 μg g(-1)). Methyl paraben was also found at high levels (>1,700 μg g(-1)) in three leave-on samples. Finally, triclosan was found at the maximum concentration limit (0.3%) laid down by the European regulation in two deodorant samples, and the total paraben concentration was close to the maximum concentration permitted (0.8%) in one leave-on sample (body milk).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Supelco
Aceton, analytical standard
Supelco
Ethylacetat, analytical standard
Supelco
Hexan, analytical standard
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Aceton, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylacetat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%