Merck
  • Graphene oxide nanosheets at the water-organic solvent interface: utilization in one-pot adsorption and reactive extraction of dye molecules.
Graphene oxide nanosheets at the water-organic solvent interface: utilization in one-pot adsorption and reactive extraction of dye molecules.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-10-11)
Ponchami Sharma, Dipankar J Borah, Manash R Das
ZUSAMMENFASSUNG
Graphene oxide (GO) is amphiphilic in nature, due to its structure, which consists of hydrophilic oxygen-containing functional groups and a hydrophobic basal plane of polyaromatic benzene rings. Due to this amphiphilicity, GO can create stable bubbles at water-organic solvent interfaces. In this study, the formation of bubbles at aqueous-organic interfaces in the presence of GO is investigated with different organic solvents. Bubble formation and transfer of GO from water to the organic phase is more prominent in aromatic solvents compared to aliphatic solvents, due to π-π interactions. Maximum transfer of GO from the aqueous to the organic phase is achieved at pH 2, and decreases with rising pH of the aqueous phase. Based on this property, and the ability of GO to adsorb cationic and anionic dye molecules, its application as a carrier for reactive extraction of cationic and anionic dye molecules is explored in toluene, kerosene, and carbon tetrachloride at pH 2 and 25 °C. The kinetics of the adsorption of the dyes onto GO nanosheets that takes place in the aqueous phase is also evaluated with different models, and a pseudo-second-order (linear) model is found to be the best fit. The adsorption isotherm data are also analyzed with different isotherm models. The electrostatic interaction and π-π interaction between the dye molecules and GO nanosheets leads to dye extraction of up to 98.2% using this technique. The dye extraction is maximum in toluene and at low dye concentration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Kaliumbromid, ≥99% trace metals basis
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Kaliumpermanganat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Kaliumbromid, ACS reagent, ≥99.0%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Natronlauge, 5.0 M
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Sigma-Aldrich
Kaliumpermanganat, ACS reagent, ≥99.0%, low in mercury
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes