Merck
  • Perturbing the cellular levels of steroid receptor coactivator-2 impairs murine endometrial function.
Perturbing the cellular levels of steroid receptor coactivator-2 impairs murine endometrial function.
PloS one (2014-06-07)
Maria M Szwarc, Ramakrishna Kommagani, Jae-Wook Jeong, San-Pin Wu, Sophia Y Tsai, Ming-Jer Tsai, Bert W O'Malley, Francesco J DeMayo, John P Lydon
ZUSAMMENFASSUNG
As pleiotropic coregulators, members of the p160/steroid receptor coactivator (SRC) family control a broad spectrum of transcriptional responses that underpin a diverse array of physiological and pathophysiological processes. Because of their potent coregulator properties, strict controls on SRC expression levels are required to maintain normal tissue functionality. Accordingly, an unwarranted increase in the cellular levels of SRC members has been causally linked to the initiation and/or progression of a number of clinical disorders. Although knockout mouse models have underscored the critical non-redundant roles for each SRC member in vivo, there are surprisingly few mouse models that have been engineered to overexpress SRCs. This deficiency is significant since SRC involvement in many of these disorders is based on unscheduled increases in the levels (rather than the absence) of SRC expression. To address this deficiency, we used recent mouse technology that allows for the targeted expression of human SRC-2 in cells which express the progesterone receptor. Through cre-loxP recombination driven by the endogenous progesterone receptor promoter, a marked elevation in expression levels of human SRC-2 was achieved in endometrial cells that are positive for the progesterone receptor. As a result of this increase in coregulator expression, female mice are severely subfertile due to a dysfunctional uterus, which exhibits a hypersensitivity to estrogen exposure. Our findings strongly support the proposal from clinical observations that increased levels of SRC-2 are causal for a number of endometrial disorders which compromise fertility. Future studies will use this mouse model to decipher the molecular mechanisms that underpin the endometrial defect. We believe such mechanistic insight may provide new molecular descriptors for diagnosis, prognosis, and/or therapy in the clinical management of female infertility.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
o-Xylen, reagent grade, ≥98.0%
Sigma-Aldrich
Progesteron, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Progesteron, ≥99%
Sigma-Aldrich
o-Xylen, anhydrous, 97%
Supelco
Toluol, analytical standard
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesteron, meets USP testing specifications
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
o-Xylen, analytical standard
Sigma-Aldrich
Progesteron, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesteron, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
USP
Progesteron, United States Pharmacopeia (USP) Reference Standard
Supelco
o-Xylen, Pharmaceutical Secondary Standard; Certified Reference Material
Progesteron für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Progesteron, European Pharmacopoeia (EP) Reference Standard
Progesteron für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Supelco
Progesteron, VETRANAL®, analytical standard