Merck
  • A pre-clinical functional assessment of an acellular scaffold intended for the treatment of hard-to-heal wounds.
A pre-clinical functional assessment of an acellular scaffold intended for the treatment of hard-to-heal wounds.
International wound journal (2013-04-06)
Gary K Shooter, Derek R Van Lonkhuyzen, Tristan I Croll, Yang Cao, Yan Xie, James A Broadbent, Dario Stupar, Emily C Lynam, Zee Upton
ZUSAMMENFASSUNG
The majority of the population experience successful wound-healing outcomes; however, 1-3% of those aged over 65 years experience delayed wound healing and wound perpetuation. These hard-to-heal wounds contain degraded and dysfunctional extracellular matrix (ECM); yet, the integrity of this structure is critical in the processes of normal wound healing. Here, we evaluated a novel synthetic matrix protein for its ability to act as an acellular scaffold that could replace dysfunctional ECM. In this regard, the synthetic protein was subjected to adsorption and diffusion assays using collagen and human dermal tissues; evaluated for its ability to influence keratinocyte and fibroblast attachment, migration and proliferation and assessed for its ability to influence in vivo wound healing in a porcine model. Critically, these experiments demonstrate that the matrix protein adsorbed to collagen and human dermal tissue but did not diffuse through human dermal tissue within a 24-hour observation period, and facilitated cell attachment, migration and proliferation. In a porcine wound-healing model, significantly smaller wound areas were observed in the test group compared with the control group following the third treatment. These data provide evidence that the synthetic matrix protein has the ability to function as an acellular scaffold for wound-healing purposes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Adenin, ≥99%
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
SAFC
L-Glutamin
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥99%
Sigma-Aldrich
Hexamethyldisiloxan, ≥98%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
SAFC
BIS-TRIS
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Adenin, BioReagent, suitable for cell culture
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥98% (TLC)