Merck
  • Synthesis and functional characterization of imbutamine analogs as histamine H3 and H4 receptor ligands.
Synthesis and functional characterization of imbutamine analogs as histamine H3 and H4 receptor ligands.
Archiv der Pharmazie (2014-02-05)
Roland Geyer, Melanie Kaske, Paul Baumeister, Armin Buschauer
ZUSAMMENFASSUNG
Imbutamine (4-(1H-imidazol-4-yl)butanamine) is a potent histamine H3 (H3R) and H4 receptor (H4R) agonist (EC50 values: 3 and 66 nM, respectively). Aiming at improved selectivity for the H4R, the imidazole ring in imbutamine was methyl-substituted or replaced by various differently substituted heterocycles (1,2,3-triazoles, 1,2,4-triazoles, pyridines, pyrimidines) as potential bioisosteres. Investigations in [(35)S]GTPγS binding assays using membranes of Sf9 insect cells expressing the respective human histamine receptor subtype revealed only very weak activity of most of the synthesized hetarylalkylamines at both receptors. By contrast, the introduction of substituents at the 4-imidazolyl ring was most effective regarding H4R selectivity. This holds for methyl substitution in position 2 and, especially, in position 5. 5-Methylimbutamine (H4R: EC50  = 59 nM, α = 0.8) was equipotent with imbutamine at the hH4R, but revealed about 16-fold selectivity for the hH4R compared to the hH3R (EC50 980 nM, α = 0.36), whereas imbutamine preferred the hH3R. The functional activities were in agreement with radioligand binding data. The results support the hypothesis that, by analogy with histamine, methyl substitution in histamine homologs offers a way to shift the selectivity in favor of the H4R.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Histamin, ≥97.0%
Supelco
Histamin, analytical standard