Merck
  • Cysteine substitution of transmembrane domain amino acids alters the ethanol inhibition of GluN1/GluN2A N-methyl-D-aspartate receptors.
Cysteine substitution of transmembrane domain amino acids alters the ethanol inhibition of GluN1/GluN2A N-methyl-D-aspartate receptors.
The Journal of pharmacology and experimental therapeutics (2015-01-31)
Minfu Xu, C Thetford Smothers, John J Woodward
ZUSAMMENFASSUNG
N-Methyl-d-aspartate receptors (NMDARs) are inhibited by behaviorally relevant concentrations of ethanol, and residues within transmembrane (TM) domains of NMDARs, including TM3 GluN1 phenylalanine 639 (F639), regulate this sensitivity. In the present study, we used cysteine (C) mutagenesis to determine whether there are additional residues within nearby TM domains that regulate ethanol inhibition on NMDARs. GluN1(F639C)/GluN2A receptors were less inhibited by ethanol than wild-type receptors, and inhibition was restored to wild-type levels following treatment with ethanol-like methanethiosulfonate reagents. Molecular modeling identified six residues in the GluN1 TM1 domain (valine V566; serine S569) and the GluN2A TM4 domain (methionine, M817; V820, F821, and leucine, L824) that were in close vicinity to the TM3 F639 residue, and these were individually mutated to cysteine and tested for ethanol inhibition and receptor function. The F639C-induced decrease in ethanol inhibition was blunted by coexpression of GluN1 TM1 mutants V566C and S569C, and statistically significant interactions were observed for ethanol inhibition among V566C, F639C, and GluN2A TM4 mutants V820C and F821C and S569C, F639C, and GluN2A TM4 mutants F821C and L824C. Ethanol inhibition was also reduced when either GluN1 TM1 mutant V566C or S569C was combined with GluN2A V820C, suggesting a novel TM1:TM4 intrasubunit site of action for ethanol. Cysteines substituted at TM3 and TM4 sites previously suggested to interact with ethanol had less dramatic effects on ethanol inhibition. Overall, the results from these studies suggest that interactions among TM1, TM3, and TM4 amino acids in NMDARs are important determinants of ethanol action at these receptors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumfluorid, ACS reagent, ≥99%
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, Molecular Biology, ≥97.0%