Merck
  • Combined exposure to bacteria and cigarette smoke resembles characteristic phenotypes of human COPD in a murine disease model.
Combined exposure to bacteria and cigarette smoke resembles characteristic phenotypes of human COPD in a murine disease model.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie (2015-01-21)
Christian Herr, Gang Han, Dong Li, Thomas Tschernig, Quoc Thai Dinh, Christoph Beißwenger, Robert Bals
ZUSAMMENFASSUNG
Abundant microbial colonization is a hallmark of COPD and smoke exposure likely increases the susceptibility to colonization and infection. The aim of the present study was to characterize the pulmonary changes of a combined exposure to cigarette smoke (CS) and microbial challenge in a preclinical murine COPD model. Animals were exposed to CS for 2 weeks, 3, and 6 months. Low and high doses of heat inactivated nontypeable Haemophilus influenzae (NTHi) were administered by inhalation during the whole exposure time. Pulmonary changes were analyzed by stereology, pulmonary function tests, measurements of inflammatory cells and mediators, and histopathology. Exposure of smoke in a relatively low concentration caused COPD-like changes of pulmonary function and only little inflammation. The coadministration of low dose NTHi (ld-NTHi) augmented a macrophage dominated inflammatory profile, while high dose NTHi (hd-NTHi) induced a neutrophilic inflammatory pattern. IL-17A secretion was solely dependent on the exposure to NTHi. Also goblet cell metaplasia and the formation of lymphoid aggregates depended on exposure to bacteria. In conclusion, the combination of exposure to smoke and bacterial compounds resulted in a mouse model that resembles several aspects of human disease. Exposure to microbial structural components appears necessary to model important pathologic features of the disease and the quantity of the exposure with microorganisms has a strong effect on the phenotype.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥95% (HPLC)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥99%
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, Grade AA-1
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥98%, BioUltra, from yeast
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, purified by column chromatography, ≥99%
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%